If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf
Select a Size
| Pack Size | SKU | Availability | Price |
|---|
About This Item
grade
Molecular Biology
Quality Segment
vapor density
2.7 (vs air)
vapor pressure
0.42 mmHg ( 20 °C)
product line
BioUltra
assay
≥99.5% (GC)
form
liquid
autoignition temp.
573 °F
expl. lim.
42 %, 63 °F
technique(s)
GC/GC: suitable
impurities
Dnases, none detected, RNases, none detected, insoluble matter, passes filter test, phosphatases, none detected, proteases, none detected, ≤0.001% free acid (as CH3COOH)
evapn. residue
≤0.005%
color
colorless
refractive index
n20/D 1.479
bp
189 °C (lit.)
mp
16-19 °C (lit.)
solubility
H2O: miscible (completely)
density
1.10 g/mL (lit.)
SMILES string
CS(C)=O
cation traces
Al: ≤0.5 mg/kg, Ba: ≤0.1 mg/kg, Bi: ≤0.1 mg/kg, Ca: ≤0.5 mg/kg, Cd: ≤0.05 mg/kg, Co: ≤0.02 mg/kg, Cr: ≤0.02 mg/kg, Cu: ≤0.02 mg/kg, Fe: ≤0.5 mg/kg, K: ≤0.5 mg/kg, Li: ≤0.1 mg/kg, Mg: ≤0.1 mg/kg, Mn: ≤0.02 mg/kg, Mo: ≤0.1 mg/kg, Na: ≤1 mg/kg, Ni: ≤0.5 mg/kg, Pb: ≤0.1 mg/kg, Sr: ≤0.1 mg/kg, Zn: ≤0.1 mg/kg
λ
neat
UV absorption
λ: 280 nm Amax: 0.22
suitability
suitable for molecular biology
InChI
1S/C2H6OS/c1-4(2)3/h1-2H3
InChI key
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
General description
Application
Other Notes
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| technique(s) GC/GC: suitable | technique(s) DNA sequencing: suitable, PCR: suitable, transfection: suitable | technique(s) immunoblotting: suitable | technique(s) - |
| grade Molecular Biology | grade Molecular Biology | grade - | grade anhydrous |
| Quality Level 200 | Quality Level 200 | Quality Level 200 | Quality Level 200 |
| assay ≥99.5% (GC) | assay ≥99.9% | assay ≥99.5% | assay ≥99.9% |
| solubility H2O: miscible (completely) | solubility H2O: miscible (completely) | solubility H2O: soluble(lit.), acetone: soluble(lit.), benzene: soluble(lit.), chloroform: soluble(lit.), ethanol: soluble(lit.) | solubility H2O: miscible (completely) |
| form liquid | form liquid | form liquid | form liquid |
Still not finding the right product?
Explore all of our products under Dimethyl sulfoxide
Storage Class
10 - Combustible liquids
wgk
WGK 1
flash_point_f
188.6 °F - closed cup
flash_point_c
87 °C - closed cup
ppe
Eyeshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 667056-1G | 04061826699812 |
| 41639-100ML | 04061832089645 |
| 41639-500ML | 04061832089652 |
-
How can I determine the shelf life / expiration / retest date of this product?
1 answer-
Helpful?
-
-
How is shipping temperature determined? And how is it related to the product storage temperature?
1 answer-
Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf
Helpful?
-
-
What is Sure/Seal™ packaging?
1 answer-
The Aldrich Sure/Seal™ packaging system is useful in the handling and storage of air-sensitive reagents.
Helpful?
-
-
Dimethyl sulfoxide (DMSO) should be a liquid. Why did it arrive as a solid?
1 answer-
DMSO supercools easily, and re-melts slowly at room temperature. The melting point is 16-19 °C. The solidified product can be re-liquefied by warming to room temperature without detriment to the product.
Helpful?
-
-
What is the DMSO dissolved in?
1 answer-
DMSO is not dissolved in any solvent. It is a pure chemical compound that is a liquid at room temperature.
Helpful?
-
-
What is the Department of Transportation shipping information for this product?
1 answer-
Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.
Helpful?
-
-
What type of filter is used to sterile-filter Dimethyl sulfoxide (DMSO)?
1 answer-
DMSO can be sterile-filtered through a 0.2 micron PTFE filter. The sterility testing is per the current US Pharmacopoeia method.
Helpful?
-
-
Does DMSO have any synthetic utility?
1 answer-
DMSO has been used to oxidize thiols and disulfides to sulfonic acids, hydrolysis of epoxides, thioalkylation of phenols, and oxidation of primary alcohols and halides, and esters of primary alcohols to aldehydes.
Helpful?
-
-
What is the molarity of DMSO?
1 answer-
DMSO will have a molarity of 14.1M, based on a density of 1.1 g/mL and a molecular weight of 78.13 g/mol.
Helpful?
-
-
Is Dimethyl sulfoxide a sterile product?
1 answer-
No, this product is not considered sterile. Product Nos. D2560 and D2438 are sterile-filtered.
Helpful?
-
