Skip to Content
Merck
All Photos(1)

Key Documents

D5879

Sigma-Aldrich

Dimethyl sulfoxide

ReagentPlus®, ≥99.5%

Synonym(s):

DMSO

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2SO
CAS Number:
Molecular Weight:
78.13
Beilstein:
506008
EC Number:
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:
NACRES:
NA.21
grade:
reagent
technique(s):
immunoblotting: suitable
bp:
189 °C (lit.)
vapor pressure:
0.42 mmHg ( 20 °C)
Pricing and availability is not currently available.

grade

reagent

Quality Level

vapor density

2.7 (vs air)

vapor pressure

0.42 mmHg ( 20 °C)

product line

ReagentPlus®

Assay

≥99.5%

form

liquid

autoignition temp.

573 °F

expl. lim.

42 %, 63 °F

technique(s)

immunoblotting: suitable

impurities

≤0.1% water (Karl Fischer)

refractive index

n20/D 1.479 (lit.)

bp

189 °C (lit.)

mp

16-19 °C (lit.)

solubility

H2O: soluble(lit.)
acetone: soluble(lit.)
benzene: soluble(lit.)
chloroform: soluble(lit.)
ethanol: soluble(lit.)

density

1.10 g/mL (lit.)

SMILES string

CS(C)=O

InChI

1S/C2H6OS/c1-4(2)3/h1-2H3

InChI key

IAZDPXIOMUYVGZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dimethyl sulfoxide is an amphipathic molecule, soluble in both aqueous and organic solvents.[1] It is commonly used as an aprotic solvent[2] and also as a reagent in organic synthesis.[3]

Application

DMSO has been used as a medium to prepare a chemiluminescence reagent that can be a substitute to commercially-available enhanced chemiluminescence kits for use in immunoblotting technique.[4]

Biochem/physiol Actions

DMSO is a highly polar substance that can be used as an industrial solvent for organic and inorganic chemicals. It is also used as a protectant during cold storage of living cells.[5][6] It is also useful for the oxidation of thiols and disulfides to sulfonic acids.[7]

Packaging

Note: D5879-1L in glass bottle /D5879-1L-D in poly bottle.

Caution

Supercools easily and remelts slowly at room temperature. Solidified product can be re-liquified by warming to room temperature without detriment to the product.

Other Notes

For information on dimethyl sulfoxide miscibility, please visit the following link:
Dimethyl Sulfoxide Miscibility/Immiscibility Table

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Bouquet et al.
Molecular reproduction and development, 40(1), 110-115 (1995-01-01)
In a previous study, we have shown that the cryopreservation of mouse oocytes caused increases in the rates of degeneration and of digynic polyploid embryos, while the fertility of frozen-thawed oocytes was decreased. In this study, we have attempted to
Julia L Zehr et al.
Biology of reproduction, 72(5), 1087-1094 (2004-12-31)
This study investigated whether prenatal androgen exposure, social rank, and body weight are factors regulating pubertal development in outdoor-housed female rhesus monkeys. Subjects' mothers received injections of testosterone enanthate (20 mg/ wk), flutamide (an androgen receptor blocker, 30 mg/kg twice
Sun-A Ock et al.
Cell transplantation, 20(8), 1231-1239 (2011-02-08)
Dimethyl sulfoxide (DMSO), a commonly used cryoprotectant in cryopreservation procedures, is detrimental to viability of cells. In this view point, a comparative study was carried out to evaluate the effect of DMSO on porcine mesenchymal stem cells (pMSCs). We compared
Lowe, O.G
The Journal of Organic Chemistry, 41 (1976)
M Ruhnke et al.
Journal of clinical microbiology, 32(9), 2092-2098 (1994-09-01)
After repeated use of fluconazole for therapy of oropharyngeal candidosis, the emergence of in vitro fluconazole-resistant Candida albicans isolates (MIC, > or = 25 micrograms/ml) together with oral candidosis unresponsive to oral dosages of up to 400 mg of fluconazole

Questions

1–7 of 7 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. Is the DMSO (Dimethyl Sulfoxide) still considered usable if it was received in a frozen or solid form?

    1 answer
    1. Yes, DMSO (Dimethyl Sulfoxide) is still considered usable even if it was received in a frozen or solid form. The solidified DMSO can be easily re-liquefied by warming it to room temperature.

      Helpful?

  4. The Dimethyl sulfoxide, Product D5879,  I ordered arrived as a solid. Is the product still acceptable?

    1 answer
    1. Sigma-Aldrich ships many products, including DMSO, at ambient temperature. As a result product D5879 might arrive as a solid depending on the weather conditions during shipment. The melting point of DMSO is 16-19°C. Therefore, if the solid is allowed to warm to room temperature, it will melt without any impact on the quality of the product.

      Helpful?

  5. What is the molarity of Product D5879, Dimethyl sulfoxide?

    1 answer
    1. Pure solvents do not technically have a molarity (defined as moles of solute per liter of solution), since there is no solute distinct from the pure material. However, an effective molar concentration of 14.1M can be determined for this product, based on a density of 1.1 g/mL and a molecular weight of 78.13 g/mol.

      Helpful?

  6. My application requires a dry solvent. How can I remove water that is present in Dimethyl sulfoxide, Product D5879?

    1 answer
    1. This product has a water content specification of not more than 0.1%, which may be low enough for your application. Other DMSO products may have more water present. Molecular sieves are typically zeolite compounds that strongly adsorb water and have carefully controlled pore sizes. If you would like, dried sieves may be placed into the bulk solvent container (about 5%-10% by volume) to remove most water.  Sigma-Aldrich offers 4 Å molecular sieves (e.g. product 334308), which would be effective in removing residual water from DMSO.

      Helpful?

  7. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service