Select a Size
| Pack Size | SKU | Availability | Price |
|---|
About This Item
biological source
mouse
conjugate
unconjugated
antibody form
culture supernatant
antibody product type
primary antibodies
clone
A103, monoclonal
description
For In Vitro Diagnostic Use in Select Regions (See Chart)
form
buffered aqueous solution
species reactivity
human
packaging
vial of 0.1 mL concentrate (281M-84), vial of 0.5 mL concentrate (281M-85), bottle of 1.0 mL predilute (281M-87), vial of 1.0 mL concentrate (281M-86), bottle of 7.0 mL predilute (281M-88)
manufacturer/tradename
Cell Marque®
technique(s)
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500
isotype
IgG1
control
melanoma
shipped in
wet ice
storage temp.
2-8°C
visualization
cytoplasmic
Gene Information
human ... MLANA(2315)
General description
Physical form
Preparation Note
Analysis Note
![]() IVD | ![]() IVD | ![]() IVD | ![]() RUO |
Other Notes
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| species reactivity human | species reactivity human | species reactivity human | species reactivity human |
| conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
| biological source mouse | biological source mouse | biological source mouse | biological source mouse |
| antibody form culture supernatant | antibody form culture supernatant | antibody form purified antibody | antibody form culture supernatant |
| clone A103, monoclonal | clone M2-7C10, monoclonal | clone A103, monoclonal | clone HMB-45+A103+T311, monoclonal |
| storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C |
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 281M-85 | 0841683132837 |
| 281M-88 | 0841683132868 |
| 281M-84 | 0841683132820 |
| 281M-87 | 0841683132851 |
| 281M-86 | 0841683132844 |







