Skip to Content

281M-9

MART-1 (Melan A) (M2-7C10) Mouse Monoclonal Antibody

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
M2-7C10, monoclonal
Application:
IHC (p)
Citations:
8
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

M2-7C10, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (281M-94), vial of 0.5 mL concentrate (281M-95), bottle of 1.0 mL predilute (281M-97), vial of 1.0 mL concentrate (281M-96), bottle of 7.0 mL predilute (281M-98)

manufacturer/tradename

Cell Marque®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500

isotype

IgG2bκ

control

melanoma

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

Gene Information

human ... MLANA(2315)

General description

MART-1 (also known as Melan A) is a melanocyte differentiation antigen. It is present in melanocytes of normal skin, retina, nevi, and in more than 85% of melanomas. This antibody is very useful in establishing the diagnosis of metastatic melanomas.

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com
MART-1 Positive Control Slides, Product No. 281S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Legal Information

Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Try our Product Selector Tool to narrow your options




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library