Skip to Content
Merck
All Photos(1)

Key Documents

69413

Supelco

Sodium hydroxide solution

suitable for HPLC, LiChropur, 49.0-51.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaOH
CAS Number:
Molecular Weight:
40.00
MDL number:
UNSPSC Code:
41116105
Pricing and availability is not currently available.

vapor pressure

3 mmHg ( 37 °C)

form

solution

concentration

49.0-51.0%

technique(s)

HPLC: suitable

impurities

≤0.1% sodium carbnate (Na2CO3)

density

1.500-1.540 g/mL at 20 °C

λ

10 % in H2O

UV absorption

λ: 240 nm Amax: ≤0.17
λ: 260 nm Amax: ≤0.15
λ: 280 nm Amax: ≤0.08
λ: 300 nm Amax: ≤0.03
λ: 500 nm Amax: ≤0.02

SMILES string

[OH-].[Na+]

InChI

1S/Na.H2O/h;1H2/q+1;/p-1

InChI key

HEMHJVSKTPXQMS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium hydroxide is widely used in numerous industrial processes, including the manufacture of pulp and paper, soaps and detergents, textiles and drinking water. It is also used in the preparation of dyes, bleaches and drugs.[1]

Application

Sodium hydroxide may be used:
  • As an extraction solvent in high-performance liquid chromatography (HPLC) in combination with inductively coupled plasma mass spectrometry (ICP-MS) (HPLC ICP-MS) speciation of selenium in enriched onion leaves.[2]
  • As an analytical reagent in HPLC and HPLC coupled electrospray ionization tandem mass spectrometry (HPLC/ESI-MS/MS) determination of flavonoids and stilbenes in red wine and related biological products.[3]

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of flavonoids and stilbenes in red wine and related biological products by HPLC and HPLC-ESI-MS-MS
Stecher G, et al.
Fresenius Journal of Analytical Chemistry, 371(1), 73-80 (2001)
HPLC analysis of carbohydrates on POLYSPHER{\textregistered} CH OH columns using pulsed amperometric detection (PAD) with sodium hydroxide as post column detection reagent
Gey, MH
Fresenius Journal of Analytical Chemistry, 356, 339-343 (2000)
HPLC-ICP-MS speciation of selenium in enriched onion leaves-a potential dietary source of Se-methylselenocysteine
Wrobel K, et al.
Food Chemistry, 86(4), 617-623 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service