Skip to Content
Merck
All Photos(1)

Key Documents

57619

Supelco

Acetic acid – triethylamine solution 1:1

suitable for HPLC, 2M:2M aqueous solution, LiChropur

Synonym(s):

Triethylamine : Acetic acid 1:1 solution, Triethylammonium acetate solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOH · N(CH2CH3)3
CAS Number:
Molecular Weight:
161.24
Beilstein:
3688333
MDL number:
UNSPSC Code:
51242002
NACRES:
NA.21
Pricing and availability is not currently available.

form

liquid

technique(s)

HPLC: suitable

λ

neat

UV absorption

λ: 250 nm Amax: ≤0.1
λ: 260 nm Amax: ≤0.05
λ: 300 nm Amax: ≤0.01
λ: 500 nm Amax: ≤0.01

SMILES string

CC(O)=O.CCN(CC)CC

InChI

1S/C6H15N.C2H4O2/c1-4-7(5-2)6-3;1-2(3)4/h4-6H2,1-3H3;1H3,(H,3,4)

InChI key

AVBGNFCMKJOFIN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Direct enantiomeric resolution of betaxolol with application to analysis of pharmaceutical products.: This study presents a method for the direct enantiomeric resolution of betaxolol, where acetic acid and triethylamine are used to facilitate the chiral separation in analytical processes, showcasing its application in pharmaceutical analysis (Hefnawy et al., 2007).

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Javier Acosta et al.
Frontiers in bioengineering and biotechnology, 8, 593-593 (2020-07-03)
Nucleoside-2'-deoxyribosyl-transferases (NDTs) catalyze a transglycosylation reaction consisting of the exchange of the 2'-deoxyribose moiety between a purine and/or pyrimidine nucleoside and a purine and/or pyrimidine base. Because NDTs are highly specific for 2'-deoxyribonucleosides they generally display poor activity on modified
Oleg V Markov et al.
Molecules (Basel, Switzerland), 25(16) (2020-08-17)
Biological activity of antisense oligonucleotides (asON), especially those with a neutral backbone, is often attenuated by poor cellular accumulation. In the present proof-of-concept study, we propose a novel delivery system for asONs which implies the delivery of modified antisense oligonucleotides
Fanghua Zhang et al.
Macromolecular bioscience, 20(7), e2000083-e2000083 (2020-06-20)
Affibody-conjugated RALA (affi-RA) are designed for delivering oligomeric 5-fluorodeoxyuridine (FUdR, metabolite of 5-FU) strand to raise the selectivity of 5-fluorouracil (5-FU), decrease its toxicity and improve its suboptimal therapeutic efficacy. The nanodrugs, FUdR@affi-RA, are spontaneously assembled by electrostatic interaction between
Nadezhda Dyrkheeva et al.
Frontiers in cell and developmental biology, 8, 604732-604732 (2021-01-12)
Human tyrosyl-DNA phosphodiesterase 1 (TDP1) belongs to the phospholipase D superfamily, whose members contain paired catalytic histidine and lysine residues within two conserved motifs and hydrolyze phosphodiester bonds. TDP1 is a DNA repair enzyme that processes 3' DNA end blocking
Zujun Liang et al.
Dalton transactions (Cambridge, England : 2003), 49(32), 11249-11259 (2020-08-07)
Platinum based anticancer agents are widely applied in clinic and their major target is believed to be DNA. Herein, the interaction of a photoactivatable diazido Pt(iv) anticancer prodrug trans,trans,trans-[Pt(N3)2(OH)2(py)2] (py = pyridine; 1) with a 15-mer single-G-containing oligodeoxynucleotide (ODN I:

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service