Skip to Content
Merck

Skip To

214701

Phosphorus pentoxide

ReagentPlus®, 99%

Synonym(s):

Phosphoric anhydride, Phosphorus(V) oxide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
P2O5
CAS Number:
Molecular Weight:
141.94
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
EC Number:
215-236-1
NACRES:
NA.21
Assay:
99%
Form:
powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor density

4.9 (vs air)

Quality Segment

vapor pressure

1 mmHg ( 384 °C), 10 mmHg ( 238 °C)

product line

ReagentPlus®

assay

99%

form

powder

pH

1.5 (20 °C, 10 g/L)

mp

340 °C (lit.)

density

2.3 g/mL at 25 °C (lit.)

cation traces

As: ≤100 mg/kg, Fe: ≤100 mg/kg, heavy metals (as Pb): ≤0.02%

SMILES string

O=P12OP3(=O)OP(=O)(O1)OP(=O)(O2)O3

InChI

1S/O10P4/c1-11-5-12(2)8-13(3,6-11)10-14(4,7-11)9-12

InChI key

YWEUIGNSBFLMFL-UHFFFAOYSA-N

General description

Phosphorus pentoxide is a deliquescent compound prepared by reacting phosphorus with air. It is a commonly used as dehydrating and condensing agent in organic synthesis.

Application

Phosphorus pentachloride has been used for the vaccum distillation of 1-methyl-2-pyrrolidinone (NMP), which was employed as mobile phase in a chromatographic analysis.

It may be used as one of the reaction components in the synthesis of dichlorine heptoxide (Cl2O7) and transition-metal phosphides (Ni2P, Co2P and MoP). P2O5 supported on alumina can be used for the solvent-free and microwave-assisted preparation of 1, 5-benzodiazepine analogs. P2O5/KX (X = Br, I) reagent system may be used for the transformation of alcohols into the corresponding alkyl iodides and bromides.
Dehydrating agent used for halogenation with tetrabutylammonium halides.

Process for Producing Halogenated Heteroaryl Compounds
Phosphorus pentoxide/methanesulfonic acid (PPMA) may be used as a condensing agent and solvent for the synthesis of:
  • High molecular weight poly(benzoxazole)s via direct polycondensation of aromatic dicarboxylic acids containing phenyl ether structure with 3,3′-dihydroxybenzidine dihydrochloride.
  • Aromatic poly(phenylene ether ether ketone)s via direct self-polycondensation of 4-(4′-phenoxyphenoxy)benzoic acids.
  • 1,3-1H-dibenzimidazole-benzene by the reaction of isophthalic acid with 1,2-diaminobenzene.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
90216029822079610
form

powder

form

powder

form

powder

form

powder, crystals or granules

assay

99%

assay

99%

assay

≥98.0%

assay

-

Quality Level

100

Quality Level

-

Quality Level

200

Quality Level

100

density

2.3 g/mL at 25 °C (lit.)

density

2.3 g/mL at 25 °C (lit.)

density

2.3 g/mL at 25 °C (lit.)

density

2.3 g/mL at 25 °C (lit.)

cation traces

As: ≤100 mg/kg, heavy metals (as Pb): ≤0.02%, Fe: ≤100 mg/kg

cation traces

As: ≤100 mg/kg, Fe: ≤100 mg/kg, heavy metals (as Pb): ≤0.02%

cation traces

NH4+: ≤0.01%, heavy metals (as Pb): ≤0.01%

cation traces

-

vapor pressure

1 mmHg ( 384 °C), 10 mmHg ( 238 °C)

vapor pressure

1 mmHg ( 384 °C), 10 mmHg ( 238 °C)

vapor pressure

1 mmHg ( 384 °C), 10 mmHg ( 238 °C)

vapor pressure

1 mmHg ( 384 °C), 10 mmHg ( 238 °C)


Still not finding the right product?

Explore all of our products under Phosphorus pentoxide


pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1A

supp_hazards

Storage Class

8A - Combustible corrosive hazardous materials

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content


Timothy Pas et al.
Molecular pharmaceutics, 17(3), 1001-1013 (2020-01-22)
In this work, a chemical (and physical) evaluation of cryogenic milling to manufacture amorphous solid dispersions (ASDs) is provided to support novel mechanistic insights in the cryomilling process. Cryogenic milling devices are considered as reactors in which both physical transitions
Synthesis of poly (benzoxazole) s by direct polycondensation of dicarboxylic acids with 3, 3??dihydroxybenzidine dihydrochloride using phosphorus pentoxide/methanesulfonic acid as condensing agent and solvent.
Ueda M, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 24(5), 1019-1026 (1986)
An efficient and selective method for the iodination and bromination of alcohols under mild conditions.
Khazdooz L, et al.
Tetrahedron Letters, 57(2), 168-171 (2016)



Global Trade Item Number

SKUGTIN
214701-2.5KG04061838773173
214701-100G04061838773159
214701-1KG04061838773166
214701-6X1KG04061837632631
214701-500G04061835276615

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters