Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Phosphorus pentoxide
Synonym(s):
Phosphoric anhydride, Phosphorus(V) oxide
Linear Formula:
P
2
O
5
CAS Number:
1314-56-3
Molecular Weight:
141.94
EC Number:
215-236-1
Products
Product Category
salts (5)
Brand
Sigma-Aldrich (5)
Supelco (1)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Special Grade
ACS reagent (2)
Anhydrous (1)
Puriss (1)
Puriss P.A. (1)
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
214701
Phosphorus pentoxide,
ReagentPlus
®
, 99%
Expand
View Pricing
298220
Phosphorus pentoxide,
powder, ACS reagent, ≥98.0%
Expand
View Pricing
902160
Phosphorus pentoxide,
anhydrous, free-flowing, Redi-Dri
™
,
ReagentPlus
®
, 99%
Expand
View Pricing
431419
Phosphorus pentoxide,
≥99.99% trace metals basis
Expand
View Pricing
79610
Phosphorus pentoxide,
desiccant, with moisture indicator
Expand
View Pricing
79609
Phosphorus pentoxide,
puriss. p.a., ACS reagent, ≥98.0% (T)
Expand
View Pricing