Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
Quality Segment
assay
≥97.5% (GC)
form
solid
autoignition temp.
480 °C
potency
178 mg/kg LD50, oral (Rat), >2000 mg/kg LD50, skin (Rat)
pH
7 (20 °C, 53 g/L in H2O)
bp
83-84 °C/1 hPa
mp
52-58 °C
transition temp
flash point 132 °C
solubility
53 g/L
density
1.13 g/cm3 at 20 °C
bulk density
530 kg/m3
storage temp.
2-30°C
SMILES string
O=Cc1c(cccc1)C=O
InChI
1S/C8H6O2/c9-5-7-3-1-2-4-8(7)6-10/h1-6H
InChI key
ZWLUXSQADUDCSB-UHFFFAOYSA-N
Application
- An investigation of o-phthaldialdehyde as a derivatization reagent and optimization of instrumental parameters for qualitative amino acid profiling via UPLC-UV Detailed examination of o-phthaldialdehyde′s efficiency in amino acid profiling to enhance diagnostic accuracies for metabolic diseases like AKU and PKU (A Aspö - 2023). Access the study.
- Correlation of the Free Amino Nitrogen and Nitrogen by O-Phthaldialdehyde Methods in the Assay of Beer This research correlates nitrogen content measurements in beer using the o-phthaldialdehyde method, discussing its implications for brewing quality control (L Otama, S Tikanoja, H Kane, S Hartikainen, L Kaski - Date not specified). Access the study.
- Turn-on fluorescence detection of glutathione based on o-phthaldialdehyde-assisted SiO2 particles Investigation into a novel fluorometric method for detecting glutathione in supplements using o-phthaldialdehyde and functionalized silica particles (P Nedeljko, M Turel, A Lobnik - 2018). Access the study.
- Biochemical Measures of Neuropathy-Glutathione Assay (GSH) A study on the biochemical assessment of neuropathy by measuring glutathione levels using o-phthaldialdehyde in a methanol solution, emphasizing the methodology′s sensitivity and reliability (E Feldman - 2019). Access the study.
Analysis Note
Melting range (lower value): ≥ 52 °C
Melting range (upper value): ≤ 56 °C
Identity (IR): passes test
1 of 1
This Item | |||
|---|---|---|---|
| assay ≥97.5% (GC) | assay - | assay ≥97% (HPLC) | assay ≥99% (HPLC) |
| Quality Level 200 | Quality Level 100 | Quality Level 200 | Quality Level 200 |
| solubility 53 g/L | solubility 53 g/L | solubility - | solubility - |
| mp 52-58 °C | mp 52-58 °C | mp 55-58 °C | mp 55-58 °C |
| density 1.13 g/cm3 at 20 °C | density 1.13 g/cm3 at 20 °C | density - | density - |
| bp 83-84 °C/1 hPa | bp 83-84 °C/1 hPa | bp - | bp - |
Still not finding the right product?
Explore all of our products under Phthaldialdehyde
signalword
Danger
Hazard Classifications
Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 3
target_organs
Respiratory system
wgk
WGK 3
flash_point_f
269.6 °F - closed cup
flash_point_c
132 °C - closed cup
hcodes
Storage Class
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.




