Skip to Content
Merck
All Photos(1)

Key Documents

P0657

Sigma-Aldrich

Phthaldialdehyde

suitable for HPLC fluorimetric detection of amino acids, ≥99% (HPLC), powder or crystals

Synonym(s):

o-Phthalaldehyde, o-Phthalic dicarboxaldehyde, Benzene-1,2-dicarboxaldehyde, OPA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H6O2
CAS Number:
Molecular Weight:
134.13
Beilstein:
878317
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.21

Quality Level

Assay

≥99% (HPLC)

form

powder or crystals

technique(s)

HPLC: suitable

color

white to light yellow

mp

55-58 °C

suitability

suitable for HPLC fluorimetric detection of amino acids

storage temp.

2-8°C

SMILES string

O=Cc1ccccc1C=O

InChI

1S/C8H6O2/c9-5-7-3-1-2-4-8(7)6-10/h1-6H

InChI key

ZWLUXSQADUDCSB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Phthaldialdehyde has been used:
  • in the preparation of O-phthaldialdehyde reagent for analysing gentamycin content
  • in the preparation of reagent for determining the degree of hydrolysis of milk proteins
  • in the measurement of free amino acids of milk samples by O-phthaldialdehyde/N-acetyl-L-cysteine (OPA/NAC) assay
  • in the derivatization of putrescine samples

Biochem/physiol Actions

Phthaldialdehyde can be used for the pre-column derivatization of amino acids for high-performance liquid chromatography (HPLC) separation. It may also be used for flow cytometric measurements of protein thiol groups.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

269.6 °F - closed cup

Flash Point(C)

132 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Colour change and proteolysis of skim milk during high pressure thermal-processing
Devi AF, et al.
Journal of Food Engineering, 147, 102-110 (2015)
Gentamicin-eluting bioresorbable composite fibers for wound healing applications
Zilberman M, et al.
Journal of Biomedical Materials Research Part A, 89(3), 654-666 (2009)
Bioactive peptides from bovine milk alpha-casein: isolation, characterization and multifunctional properties
Srinivas S and Prakash V
International Journal of Peptide Research and Therapeutics, 16(1), 7-15 (2010)
Gene expression knockdown by modulating synthetic small RNA expression in Escherichia coli
Noh M, et al.
Cell Systems, 5(4), 418-426 (2017)
Haruna Nagase et al.
Biomolecules, 10(1) (2020-01-16)
Sulfation is a common modification of extracellular glycans, tyrosine residues on proteins, and steroid hormones, and is important in a wide variety of signaling pathways. We investigated the role of sulfation on endogenous oxidative stress, such as glutamate-induced oxytosis and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service