Skip to Content
Merck

Skip To

8.02482

Chromium(II) chloride

for synthesis

Synonym(s):

Chromium(II) chloride, Chromium dichloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
Cl2Cr
CAS Number:
Molecular Weight:
122.90
UNSPSC Code:
12352302
EC Index Number:
233-163-3
NACRES:
NA.22
MDL number:
Grade:
synthesis grade
Form:
powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

synthesis grade

Quality Level

form

powder

potency

1870 mg/kg LD50, oral (Rat)

reaction suitability

reagent type: reductant

mp

823-825 °C

storage temp.

2-30°C

SMILES string

[Cr](Cl)Cl

InChI

1S/2ClH.Cr/h2*1H;/q;;+2/p-2

InChI key

XBWRJSSJWDOUSJ-UHFFFAOYSA-L

Analysis Note

Assay (oxidimetric): 89.0 - 93.0 %
Identity (ICP-OES): passes test

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8.180908.222661.00229
form

powder

form

powder

form

solid

form

solid

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

300

grade

synthesis grade

grade

synthesis grade

grade

-

grade

-

mp

823-825 °C

mp

1152 °C

mp

197 °C

mp

197 °C

potency

1870 mg/kg LD50, oral (Rat)

potency

440 mg/kg LD50, oral (Rat)

potency

-

potency

-

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

2-30°C


Still not finding the right product?

Explore all of our products under Chromium(II) chloride


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
802482001004022536373645
802482000004022536373638

Questions

Reviews

No rating value

Active Filters