![Chromium(II) chloride for synthesis](/deepweb/assets/sigmaaldrich/product/structures/332/791/9aa39bd5-dabb-41c2-8d9a-f8a2804feaaa/640/9aa39bd5-dabb-41c2-8d9a-f8a2804feaaa.png)
Chromium(II) chloride
![Chromium(II) chloride for synthesis](/deepweb/assets/sigmaaldrich/product/structures/332/791/9aa39bd5-dabb-41c2-8d9a-f8a2804feaaa/640/9aa39bd5-dabb-41c2-8d9a-f8a2804feaaa.png)
Synonym(s):
Chromium dichloride, Chromous chloride, Chromium(II) chloride
Linear Formula:
CrCl2
CAS Number:
Molecular Weight:
122.90
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Reagent Type
Core
Special Grade