Skip to Content
Merck

Skip To

324880

(–)-Epigallocatechin Gallate

One of the main polyphenolic constituents of green tea that exhibits potent antitumor, anti-inflammatory, and antioxidant properties.

Synonym(s):

(–)-Epigallocatechin Gallate, EGCG, (2R,3R)-2-(3,4,5-Trihydroxyphenyl)-3,4-dihydro-1[2H]-benzopyran-3,5,7-triol-3-(3,4,5-trihydroxybenzoate), HAT Inhibitor X, Histone Acetyltransferase Inhibitor X, p300/CBP Inhibitor VIII, PCAF Inhibitor V, DNA MTase Inhibitor IV, DNA Methyltransferase Inhibitor IV

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C22H18O11
CAS Number:
Molecular Weight:
458.37
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥95% (HPLC)
Form:
solid
Storage condition:
OK to freeze, protect from light
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

Merck USA index - 14, 3526

Quality Segment

assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, protect from light

color

faint pink

solubility

water: 5 mg/mL, DMSO: soluble

shipped in

ambient

storage temp.

−20°C

SMILES string

O1[C@@H]([C@@H](Cc4c1cc(cc4O)O)OC(=O)c3cc(c(c(c3)O)O)O)c2cc(c(c(c2)O)O)O

InChI

1S/C22H18O11/c23-10-5-12(24)11-7-18(33-22(31)9-3-15(27)20(30)16(28)4-9)21(32-17(11)6-10)8-1-13(25)19(29)14(26)2-8/h1-6,18,21,23-30H,7H2/t18-,21-/m1/s1

InChI key

WMBWREPUVVBILR-WIYYLYMNSA-N

General description

One of the main polyphenolic constituents of green tea that exhibits potent antitumor, anti-inflammatory, and antioxidant properties. Arrests cell cycling at G0/G1 phase and induces apoptosis in a dose-dependent manner. Shown to inhibit PMA-induced skin thickening and to activate of protein kinase C. Also activates ornithine decarboxylase and interleukin-1α mRNA and protein expression. Acts as an inhibitor of inducible nitric oxide synthase (iNOS) gene expression and enzyme activity. Also blocks peroxynitrite-mediated formation of 8-oxodeoxyguanosine and 3-nitrotyrosine. Strongly and directly inhibits telomerase in cell-free systems and in cancer cell lines. An inhibitor of Dnmt1 (IC50 = 210-470 nM). Acts as a selective and noncompetitive inhibitor of HAT activities (IC50 = 30, 50, 60 and 70 µM for p300, CBP, PCAF and TIP60) with minimal effects on HDAC, SIRT and HMTase.

Biochem/physiol Actions

Primary Target
PMA-induced skin thickening
Product does not compete with ATP.
Reversible: no
Target IC50: 210-470 nM against Dnmt1; 30, 50, 60 and 70 µM for p300, CBP, PCAF and TIP60
Cell permeable: no

Packaging

Packaged under inert gas

Preparation Note

Following reconstitution, aliquot and freeze at -20°C. DMSO stock solutions are stable for up to 3 months at -20°C.

Other Notes

Choi, K.C., et al. 2009. Cancer Res.69, 583.
Dell′Aicia, I., et al. 2004. EMBO reports 5, 1.
Tachibana, H., et al. 2004. Nat. Struct. Mol. Biol.11, 380.
Naasami, I., et al. 1998. Biochem. Biophys. Res. Commun. 249, 391.
Ahmad, N., et al. 1997. J. Natl. Cancer89, 1881.
Chan, M.M.-Y., et al. 1997. Biochem. Pharmacol. 54, 1281.
Lin, Y.-L., and Lin, J.-K. 1997. Mol. Pharmacol. 52, 465.
Fiala, E.S., et al. 1996. Experientia 52, 922.
Katiyar, S.K., et al. 1995. J. Invest. Dermatol. 105, 394.
Liao, S., et al. 1995. Cancer Lett. 96, 239.
Yamane, T., et al. 1995. Cancer Res. 55, 2081.
Huang, M.T., et al. 1992. Carcinogenesis 13, 947.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Irritant (B)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
E426893894E4143
form

solid

form

-

form

-

form

-

assay

≥95% (HPLC)

assay

≥80% (HPLC)

assay

≥98.0% (HPLC)

assay

≥95%

manufacturer/tradename

Calbiochem®

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

-

Quality Level

100

Quality Level

200

Quality Level

100

Quality Level

200

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage condition

OK to freeze, protect from light

storage condition

-

storage condition

-

storage condition

-


Still not finding the right product?

Explore all of our products under (–)-Epigallocatechin Gallate


pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters