Skip to Content
Merck

Skip To

E4143

(−)-Epigallocatechin gallate

≥95%

Synonym(s):

(−)-cis-2-(3,4,5-Trihydroxyphenyl)-3,4-dihydro-1(2H)-benzopyran-3,5,7-triol 3-gallate, (−)-cis-3,3′,4′,5,5′,7-Hexahydroxy-flavane-3-gallate, EGCG

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C22H18O11
CAS Number:
Molecular Weight:
458.37
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12352205
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

assay

≥95%

solubility

H2O: ≥5 mg/mL, clear

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

2-8°C

SMILES string

Oc1cc(O)c2C[C@@H](OC(=O)c3cc(O)c(O)c(O)c3)[C@H](Oc2c1)c4cc(O)c(O)c(O)c4

InChI

1S/C22H18O11/c23-10-5-12(24)11-7-18(33-22(31)9-3-15(27)20(30)16(28)4-9)21(32-17(11)6-10)8-1-13(25)19(29)14(26)2-8/h1-6,18,21,23-30H,7H2/t18-,21-/m1/s1

InChI key

WMBWREPUVVBILR-WIYYLYMNSA-N

Gene Information

human ... CYP1A2(1544)

General description

(−)-Epigallocatechin gallate from green tea is the prime bioactive component, accounting for 50-80% of the total catechin content.[1] It comprises a chemical structure similar to that of epicatechin gallate (ECG), an ester of gallic acid and epigallocatechin.[2]

Application

(-)-Epigallocatechin gallate has been used:
  • as an anti-tumor agent on murine TRAMP metastatic prostate cell line by cell proliferation assay, apoptosis detection and modified Boyden-chamber assay
  • induces cell death in acute myeloid leukaemia through death associated protein kinase-2 pathway analyzed through cell viability, cell cycle and apoptosis assay
  • as an inhibitor of osteoclast differentiation in murine preosteoclast cell line RAW264.7
  • to promote myogenic differentiation

Biochem/physiol Actions

(-)-Epigallocatechin gallate (EGCG), an antioxidant polyphenol flavonoid exerts anti-tumor properties by inhibiting telomerase and DNA methyltransferase activity.[1] EGCG inhibits the expression of matrix metalloproteinase-2 (MMP-2), MMP-9 and reduces the invasiveness. EGCG blocks the activation of epidermal growth factor (EGF) receptors and human epidermal growth factor receptor-2 (HER-2).[1] EGCG increases bone mineral density and reduces bone resorption. EGCG inhibits osteoclastogenesis by inhibiting receptor activator of nuclear factor κ-B ligand (RANKL) induced nuclear factor κ B (NF-κB) transcriptional activity. EGCG reduces skeletal muscle atrophy. EGCG has anti-aging property and increases myogenic differentiation. EGCG inhibits fatty acid synthase and glutamate dehydrogenase activity.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
E426893894PHR1333
assay

≥95%

assay

≥80% (HPLC)

assay

≥98.0% (HPLC)

assay

-

Quality Level

200

Quality Level

200

Quality Level

100

Quality Level

300

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

application(s)

food and beverages

application(s)

pharmaceutical (small molecule)

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

solubility

H2O: ≥5 mg/mL, clear

solubility

-

solubility

-

solubility

-

Gene Information

human ... CYP1A2(1544)

Gene Information

human ... CYP1A2(1544)

Gene Information

-

Gene Information

-


Still not finding the right product?

Explore all of our products under (−)-Epigallocatechin gallate


pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Sens. 1

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



(-)-Epigallocatechin gallate inhibition of osteoclastic differentiation via NF-kappaB
Lin RW, et al.
Biochemical and Biophysical Research Communications, 379(4), 1033-1037 (2009)
(-)-Epigallocatechin-3-gallate stimulates myogenic differentiation through TAZ activation
Kim AR, et al.
Biochemical and Biophysical Research Communications, 486(2), 378-384 (2017)
Takuya Miyagawa et al.
Pharmaceutics, 12(5) (2020-05-02)
Neovascularization (NV) of the cornea disrupts vision which leads to blindness. Investigation of antiangiogenic, slow-release and biocompatible approaches for treating corneal NV is of great importance. We designed an eye drop formulation containing gelatin/epigallocatechin-3-gallate (EGCG) nanoparticles (NPs) for targeted therapy



Global Trade Item Number

SKUGTIN
E4143-50MG04061835544455

Questions

1–9 of 9 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. What is the solution stability of (-)-Epigallocatechin gallate (E4143)?

    1 answer
    1. A solution of (-)-Epigallocatechin gallate in water at a concentration of 0.3 mg/mL was found to have significantly decreased in purity (from 99.6% to 81.7%) after 2.5 hours at room temperature. A similar solution of (-)-Epigallocatechin gallate in water at a concentration of 0.3 mg/mL was found to have decreased in purity only slightly (from 99.5% to 99.3%) after 2.0 hours at 4°C.

      Helpful?

  4. Is (-)-Epigallocatechin gallate (Product No. E4143) for human use

    1 answer
    1. No. Product No. E4143 ((-)-Epigallocatechin gallate) is for laboratory research use only, not for human use.

      Helpful?

  5. How does the storage temperature relate to shipping conditions?

    1 answer
    1. The storage conditions that a Sigma-Aldrich catalog and label recommend for products are deliberately conservative. For many products, long-term storage at low temperatures will increase the time during which they are expected to remain in specification and therefore are labeled accordingly. Where short-term storage, shipping time frame, or exposure to conditions other than those recommended for long-term storage will not affect product quality, Sigma-Aldrich will ship at ambient temperature. The products sensitive to short-term exposure to conditions other than their recommended long-term storage are shipped on wet or dry ice. Ambient temperature shipping helps to control shipping costs for our customers. At any time, our customers can request wet- or dry-ice shipment, but the special handling is at customer expense if our product history indicates that the product is stable for regular shipment.

      Helpful?

  6. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  7. Is (-)-Epigallocatechin gallate (Product No. E4143) sterile and tested for endotoxins?

    1 answer
    1. Product No. E4143 ((-)-Epigallocatechin gallate) is not prepared under sterile conditions and has not been tested for endotoxins.

      Helpful?

  8. Can (-)-Epigallocatechin gallate be determined in plasma and urine?

    1 answer
    1. The determination of (-)-Epigallocatechin gallate in plasma and urine by HPLC has been reported in Cancer Epidemiol. Biomark. Prev., 4, 393 (1995).

      Helpful?

  9. What is the solubility of (-)-Epigallocatechin gallate (E4143)?

    1 answer
    1. We have tested the solubility of (-)-Epigallocatechin gallate at 5 mg/mL in water.

      Helpful?

Reviews

No rating value

Active Filters