Skip to Content
Merck

Skip To

1.08372

Triethanolamine

(Trolamine)

Synonym(s):

Triethanolamine, Tris(2-hydroxyethyl)amine, 2,2′,2′′-Trihydroxytriethylamine, TEA, 2,2′,2′′-Nitrilotriethanol, Tris(2-hydroxyethyl)amine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
(HOCH2CH2)3N
CAS Number:
Molecular Weight:
149.19
EC Index Number:
203-049-8
UNSPSC Code:
12352104
Beilstein/REAXYS Number:
1699263
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor density

5.14 (vs air)

Quality Level

vapor pressure

<0.01 hPa ( 20 °C), 0.01 mmHg ( 20 °C)

assay

99.0-103.0% anhydrous basis (acidimetric)

form

liquid

autoignition temp.

325 °C (DIN 51794), 600 °F

expl. lim.

3.6-7.2 % (v/v), 8.5 %

refractive index

n20/D 1.485 (lit.)

pH

10.5 (20 °C, 15 g/L in H2O)

useful pH range

7.3-8.3

pKa (25 °C)

7.8

bp

190-193 °C/5 mmHg (lit.), 360 °C/1013 hPa (decomposition)

mp

17.9-21 °C (lit.), 21 °C

transition temp

flash point 190 °C

density

1.12 g/cm3 at 20 °C, 1.124 g/mL at 25 °C (lit.)

application(s)

pharma/biopharma processes

storage temp.

15-25°C

SMILES string

OCCN(CCO)CCO

InChI

1S/C6H15NO3/c8-4-1-7(2-5-9)3-6-10/h8-10H,1-6H2

InChI key

GSEJCLTVZPLZKY-UHFFFAOYSA-N

General description

We offer a range of high quality critical raw materials plus the documentation and expertise to help our customers meet stringent regulatory requirements. Our SAFC® portfolio of high-quality products withstands strict quality control procedures.

Application

Triethanolamine (Trolamine) is a tertiary amine that acts as a basic buffer component, typically used as a buffer in biopharmaceutical processes, such as cell lysis and chromatography.

Legal Information

SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.3714850926T58300
form

liquid

form

liquid

form

-

form

viscous liquid

assay

99.0-103.0% anhydrous basis (acidimetric)

assay

-

assay

≥99.0% (GC)

assay

98%

application(s)

pharma/biopharma processes

application(s)

liquid formulation
pharmaceutical
semi-solid formulation

application(s)

cleaning products
cosmetics
food and beverages
personal care

application(s)

-

useful pH range

7.3-8.3

useful pH range

7.3-8.3

useful pH range

7.3-8.3

useful pH range

7.3-8.3

expl. lim.

3.6-7.2 % (v/v), 8.5 %

expl. lim.

8.5 %

expl. lim.

8.5 %

expl. lim.

8.5 %

storage temp.

15-25°C

storage temp.

2-25°C

storage temp.

-

storage temp.

-


Still not finding the right product?

Explore all of our products under Triethanolamine


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

354.2 °F - closed cup

flash_point_c

179 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library





Global Trade Item Number

SKUGTIN
124036-25G04061838722713
124036-100G04061838345578
108372100004022536925813
108372602704054839520822
108372250004027269130152
108372602504027269244866
108372618004027269244880

Questions

Reviews

No rating value

Active Filters