Skip to Content
Merck
All Photos(1)

Key Documents

T58300

Sigma-Aldrich

Triethanolamine

reagent grade, 98%

Synonym(s):

2,2′,2′′-Nitrilotriethanol, Tris(2-hydroxyethyl)amine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HOCH2CH2)3N
CAS Number:
Molecular Weight:
149.19
Beilstein:
1699263
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

grade

reagent grade

Quality Level

vapor density

5.14 (vs air)

vapor pressure

0.01 mmHg ( 20 °C)

Assay

98%

form

viscous liquid

autoignition temp.

600 °F

expl. lim.

8.5 %

refractive index

n20/D 1.485 (lit.)

useful pH range

7.3-8.3

pKa (25 °C)

7.8

bp

190-193 °C/5 mmHg (lit.)

mp

17.9-21 °C (lit.)

solubility

soluble 149 g/L at 20 °C (completely)

density

1.124 g/mL at 25 °C (lit.)

functional group

amine
hydroxyl

SMILES string

OCCN(CCO)CCO

InChI

1S/C6H15NO3/c8-4-1-7(2-5-9)3-6-10/h8-10H,1-6H2

InChI key

GSEJCLTVZPLZKY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Triethanolamine (TEA) is a quaternary ammonium compound commonly used as a buffer and emulsifier.[1]

Application

Triethanolamine can be used as:
  • A nitrogen precursor in the preparation of carbon dot derived nanomaterials.[2]
  • A cross-linking agent in the synthesis of methylaluminoxane pre-catalysts applicable in the polymerization of ethylene.[3]
  • A reducing agent to synthesize Ag nanoparticles of biological importance.[4]

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

354.2 °F - closed cup

Flash Point(C)

179 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carbonization conditions influence the emission characteristics and the stability against photobleaching of nitrogen doped carbon dots
Xiong Y, et al.
Nanoscale, 9(32), 11730-11738 (2017)
Fabrication of an effective avermectin nanoemulsion using a cleavable succinic ester emulsifier
W Guan, et al.
Journal of Agricultural and Food Chemistry, 19, 74-76 (2018)
Preparation of Ag nanoparticles with triethanolamine as reducing agent and their antibacterial property
Jia Z, et al.
Colloids and Surfaces. A, Physicochemical and Engineering Aspects, 419, 174-179 (2013)
Spherical MgCl2-supported MAO pre-catalysts: preparation, characterization and activity in ethylene polymerization
Guan Z, et al.
J. Mol. Catal. A: Chem., 188(1-2), 123-131 (2002)
Friederike Saaber et al.
Cell reports, 26(6), 1473-1488 (2019-02-07)
Phosphorylation of heptahelical receptors is thought to regulate G protein signaling, receptor endocytosis, and non-canonical signaling via recruitment of β-arrestins. We investigated chemokine receptor functionality under phosphorylation-deficient and β-arrestin-deficient conditions by studying interneuron migration in the embryonic cortex. This process

Questions

1–9 of 9 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. Is this a product solid form or in liquid form ??

    1 answer
    1. As mentioned in the 'PROPERTIES' section, this product is supplied as a viscous liquid. The melting point range is 17.9 - 21 °C. The product may solidify at lower temperatures. Warming the material to reliquify will not impact the integrity of the material.

      Helpful?

  4. Why was the product T58300 Triethanolamine received in a solid state when it is supposed to be a liquid?

    1 answer
    1. Triethanolamine has a low melting point of 17.9 - 21°C, and it is a viscous liquid above 22 degrees Celsius. During the coldest months of the year, if the atmospheric temperature is below 21 degrees Celsius, this liquid product will arrive in a solid state. To remedy this, place the bottle in a water bath above 22 degrees Celsius to melt the product. The quality of the product will not be affected by status changes.

      Helpful?

  5. What is the boiling point of Product T58300, Triethanolamine?

    1 answer
    1. The boiling point is not determined by Sigma-Aldrich, but is reported to be 190-193°C at a reduced pressure of 5mm Hg.

      Helpful?

  6. Product T58300, Triethanolamine, was received as a solid and it usually arrives as a liquid. Is the product still acceptable?

    1 answer
    1. Yes, the product is still acceptable. Triethanolamine has a melting point of 17.9-21°C, which is near room temperature. The item ships under ambient conditions. As a result, it may arrive as a solid when exposed to cold temperatures (especially during the winter months). Gentle heating in a warm water bath will easily convert the material back into its expected viscous liquid appearance.

      Helpful?

  7. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  8. How can I dilute Product T58300, Triethanolamine?

    1 answer
    1. According to the chemicals encyclopedia published by the Royal Society of Chemistry (which we also offer as product Z705322), triethanolamine is expected to be miscible with water, methanol or acetone.

      Helpful?

  9. What kind of gloves would be recommended when working with Product T58300, Triethanolamine?

    1 answer
    1. When working with the material, we would recommend neoprene, nitrile or butyl gloves. For example, the Best® butyl, ChemMaster™ or N-DEX®  gloves.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service