Skip to Content
Merck

Skip To

D-122

Diethylpropion-D10 hydrochloride solution

100 μg/mL in methanol (with 1% 1 M HCl, (as free base)), certified reference material, ampule of 1 mL, Cerilliant®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C13H9D10NO · HCl
CAS Number:
Molecular Weight:
251.82
UNSPSC Code:
41116107
NACRES:
NA.24
EC Number:
200-659-6
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol (with 1% 1 M HCl, (as free base))

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

O=C(C(C)N(C([2H])(C([2H])([2H])[2H])[2H])C([2H])(C([2H])([2H])[2H])[2H])C1=CC=CC=C1.[H]Cl

InChI

1S/C13H19NO.ClH/c1-4-14(5-2)11(3)13(15)12-9-7-6-8-10-12;/h6-11H,4-5H2,1-3H3;1H/i1D3,2D3,4D2,5D2;

InChI key

ICFXZZFWRWNZMA-IDHUOJFFSA-N

General description

Also known as diethylcathinone or amfepramone, diethylpropion is a stimulant and appetite suppressant drug of the phenethylamine, amphetamine, and cathinone chemical classes. This new internal standard is suitable for numerous GC/MS and LC/MS applications from forensic analysis and clinical toxicology testing to urine drug testing and isotope dilution methods

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Supply conditions do not apply to the regions and states of Brazil: North, Northeast, Mato Grosso do Sul, Mato Grosso, and Rio Grande do Sul.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
D-121D-161G-017
grade

certified reference material

grade

certified reference material

grade

certified reference material

grade

certified reference material

format

single component solution

format

single component solution

format

single component solution

format

single component solution

form

liquid

form

liquid

form

liquid

form

liquid

concentration

100 μg/mL in methanol (with 1% 1 M HCl, (as free base))

concentration

1.0 mg/mL in methanol (with 1% 1 M HCl; as free base)

concentration

1 mg/mL in methanol (as free base)

concentration

1.0 mg/mL in methanol

Quality Level

300

Quality Level

300

Quality Level

300

Quality Level

300

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Lili Aslostovar et al.
Cell reports. Medicine, 2(2), 100202-100202 (2021-03-06)
The aberrant expression of dopamine receptors (DRDs) in acute myeloid leukemia (AML) cells has encouraged the repurposing of DRD antagonists such as thioridazine (TDZ) as anti-leukemic agents. Here, we access patient cells from a Phase I dose escalation trial to



Global Trade Item Number

SKUGTIN
S-056-1ML04061836829612
D-122-1ML04061833557259
HS23452104061835302277

Questions

Reviews

No rating value

Active Filters