Skip to Content
Merck

Skip To

D-121

Diethylpropion hydrochloride solution

1.0 mg/mL in methanol (with 1% 1 M HCl; as free base), certified reference material, Cerilliant®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C13H20ClNO
CAS Number:
Molecular Weight:
241.76
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule B (Switzerland); Pszichotróp anyag / Psychotropic Substance (Hungary), 78/2022. (XII. 28.) BM rendelet, kontrollierte Droge in Deutschland

concentration

1.0 mg/mL in methanol (with 1% 1 M HCl; as free base)

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

Cl.CCN(CC)C(C)C(=O)c1ccccc1

InChI

1S/C13H19NO.ClH/c1-4-14(5-2)11(3)13(15)12-9-7-6-8-10-12;/h6-11H,4-5H2,1-3H3;1H

InChI key

ICFXZZFWRWNZMA-UHFFFAOYSA-N

Gene Information

General description

Also known as diethylcathinone or amfepramone, diethylpropion is a stimulant and appetite suppressant drug of the phenethylamine, amphetamine, and cathinone chemical classes. This reference standard is applicable for use in urine drug testing, clinical toxicology, or forensic analysis by LC-MS/MS or GC/MS.

Legal Information

German
Dieses Produkt fällt unter das Betäubungsmittelgesetz (BtMG). Für eine Bestellung dieses Produktes ist eine Erlaubnis nach § 3 BtMG zwingend erforderlich, es sei denn, es greift eine Ausnahme von der Erlaubnispflicht nach § 4 oder § 26 BtMG.

English
This product is subject to the German Narcotics Act. A permit under Section 3 of the German Narcotics Act is mandatory for ordering this product unless an exemption from the permit requirement under Section 4 or Section 26 of the German Narcotics Act applies.
CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
D-122D-161V-902
grade

certified reference material

grade

certified reference material

grade

certified reference material

grade

certified reference material

format

single component solution

format

single component solution

format

single component solution

format

single component solution

feature

Snap-N-Spike®/Snap-N-Shoot®

feature

Snap-N-Spike®/Snap-N-Shoot®

feature

Snap-N-Spike®/Snap-N-Shoot®

feature

Snap-N-Spike®/Snap-N-Shoot®

concentration

1.0 mg/mL in methanol (with 1% 1 M HCl; as free base)

concentration

100 μg/mL in methanol (with 1% 1 M HCl, (as free base))

concentration

1 mg/mL in methanol (as free base)

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

form

liquid

form

liquid

form

liquid

form

liquid


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
D-121-1ML04061833557242
DWKW348924-J-1EA04065273636598

Questions

Reviews

No rating value

Active Filters