Skip to Content
Merck

Skip To

1045337

USP

Avobenzone

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

1-(4-Methoxyphenyl)-3-(4-tert-butylphenyl)-1,3-propanedione, Butyl methoxydibenzoylmethane

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C20H22O3
CAS Number:
Molecular Weight:
310.39
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

avobenzone

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

COc1ccc(cc1)C(=O)CC(=O)c2ccc(cc2)C(C)(C)C

InChI

1S/C20H22O3/c1-20(2,3)16-9-5-14(6-10-16)18(21)13-19(22)15-7-11-17(23-4)12-8-15/h5-12H,13H2,1-4H3

InChI key

XNEFYCZVKIDDMS-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Avobenzone USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
16633PHR10731011721
grade

pharmaceutical primary standard

grade

analytical standard

grade

certified reference material, pharmaceutical secondary standard

grade

pharmaceutical primary standard

manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

USP

format

neat

format

neat

format

neat

format

neat

application(s)

pharmaceutical (small molecule)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

application(s)

environmental
pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

API family

avobenzone

API family

-

API family

avobenzone

API family

adapalene


Still not finding the right product?

Explore all of our products under Avobenzone


hcodes

Hazard Classifications

Aquatic Chronic 4

Storage Class

11 - Combustible Solids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Ok-Nam Bae et al.
Toxicology and applied pharmacology, 283(2), 147-155 (2015-01-27)
Allergic contact dermatitis (ACD) is a cell-mediated immune response that involves skin sensitization in response to contact with various allergens. Angiogenesis and lymphangiogenesis both play roles in the allergic sensitization process. Epidermal keratinocytes can produce vascular endothelial growth factor (VEGF)
Chi-Feng Hung et al.
International journal of pharmaceutics, 471(1-2), 135-145 (2014-05-27)
Intrinsic aging and photoaging modify skin structure and components, which subsequently change percutaneous absorption of topically applied permeants. The purpose of this study was to systematically evaluate drug/sunscreen permeation via young and senescent skin irradiated by ultraviolet (UV) light. Both
Marina G Pintado-Herrera et al.
Analytica chimica acta, 851, 1-13 (2014-12-03)
This work presents the development, optimization and validation of a multi-residue method for the simultaneous determination of 102 contaminants, including fragrances, UV filters, repellents, endocrine disruptors, biocides, polycyclic aromatic hydrocarbons (PAHs), polychlorinated biphenyls (PCBs), and several types of pesticides in



Global Trade Item Number

SKUGTIN
1045337-500MG04061838671905

Questions

Reviews

No rating value

Active Filters