Skip to Content
Merck
All Photos(1)

Key Documents

PHR1073

Supelco

Avobenzone

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

1-(4-Methoxyphenyl)-3-(4-tert-butylphenyl)-1,3-propanedione, Butyl methoxydibenzoylmethane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H22O3
CAS Number:
Molecular Weight:
310.39
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1045337

API family

avobenzone

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental
pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

COc1ccc(cc1)C(=O)CC(=O)c2ccc(cc2)C(C)(C)C

InChI

1S/C20H22O3/c1-20(2,3)16-9-5-14(6-10-16)18(21)13-19(22)15-7-11-17(23-4)12-8-15/h5-12H,13H2,1-4H3

InChI key

XNEFYCZVKIDDMS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Avobenzone is an active sunscreen ingredient, used in sunscreen lotions and cosmetic formulations.
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Avobenzone may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations using reversed-phase high-performance liquid chromatography technique.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAB3693 in the slot below. This is an example certificate only and may not be the lot that you receive.

related product

Product No.
Description
Pricing

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Woan-Ruoh Lee et al.
Toxicology letters, 211(2), 150-158 (2012-04-10)
While laser skin resurfacing is expected to result in reduced barrier function and increased risk of drug absorption, the extent of the increment has not yet been systematically investigated. We aimed to establish the skin permeation profiles of tetracycline and
Kerry M Hanson et al.
Journal of cosmetic science, 54(6), 589-598 (2004-01-20)
The ability of sunscreens and antioxidants to deactivate highly destructive reactive oxygen species in human skin has remained inconclusive. Two-photon fluorescence imaging microscopy was used to determine the effect of sunscreen/antioxidant combinations upon UV-induced ROS generation in ex vivo human
Daniele Dondi et al.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology, 5(9), 835-843 (2006-10-19)
A systematic investigation of two well-known and popular commercial suncreams reveals significant degradation when exposed to simulated UV sunlight at an irradiance corresponding to natural sunlight. We have examined the photochemistry of two widely used sunscreen active agents in pure
Robert M Sayre et al.
Photochemistry and photobiology, 81(2), 452-456 (2004-11-25)
A major concern raised about photostability studies of sunscreen products is that the photodegradation of sunscreens does not readily translate into changes in product performance. This study examines the correlation between photochemical degradation of sunscreen agents and changes in protection
Serge Forestier
Journal of the American Academy of Dermatology, 58(5 Suppl 2), S133-S138 (2008-04-25)
The risks associated with cumulative or overexposure to ultraviolet (UV) radiation are now well identified. They involve both UVB (290-320 nm) and UVA (320-400 nm) components. UVB radiation is still considered to be the major factor responsible for most harmful

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service