Skip to Content
Merck

Skip To

M5904

Mineral oil

BioReagent, Molecular Biology, light oil

Synonym(s):

light mineral oil

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12181504
NACRES:
NA.52
EC Number:
232-455-8
MDL number:
Grade:
Molecular Biology
Technique(s):
PCR: suitable
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

Molecular Biology

Quality Segment

product line

BioReagent

form

light oil

technique(s)

PCR: suitable

color

colorless

refractive index

n20/D 1.467 (lit.)

density

0.84 g/mL at 25 °C (lit.)

suitability

suitable for overlaying aqueous reactions and centrifuge gradients

foreign activity

DNase, RNase, Protease, none detected

storage temp.

room temp

InChI key

AEOVEGJBKQQFOP-DDVLFWKVSA-L

General description

Mineral oils are usually seen as a mixture of liquid hydrocarbons. It is derived from crude oil by distillation and refining.

Application

Mineral oil has been used:
  • to suspend the gels for capillary contraction assay
  • in polymerase chain reaction (PCR) reactions mixture to form a vapor barrier in order to prevent the evaporation of the reaction at elevated temperatures
  • as a continuous phase to prevent droplet coalescence in emulsification system

Mineral oil is suitable for use as an overlay to control evaporation and cross contamination of samples in a variety of molecular biology applications.[1][2][3]

Biochem/physiol Actions

Mineral oil delivers a better insulation strength, when compared to air. It enhances the elimination of heat from the windings, when used in a power transformer.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
M5310M8662330760
Quality Level

200

Quality Level

300

Quality Level

100

Quality Level

200

grade

Molecular Biology

grade

-

grade

-

grade

-

technique(s)

PCR: suitable

technique(s)

cell culture | embryo: suitable

technique(s)

PCR: suitable

technique(s)

-

form

light oil

form

light oil

form

liquid

form

liquid (colorless)

density

0.84 g/mL at 25 °C (lit.)

density

0.82-0.88 g/mL at 25 °C

density

0.84 g/mL at 25 °C (lit.)

density

0.862 g/mL at 25 °C

refractive index

n20/D 1.467 (lit.)

refractive index

n20/D 1.467 (lit.)

refractive index

n20/D 1.467 (lit.)

refractive index

n20/D 1.467 (lit.)


Still not finding the right product?

Explore all of our products under Mineral oil


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

No data available

flash_point_c

No data available

ppe

Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Process Fluids for Abrasive Machining
Tribology of Abrasive Machining Processes, 531-585 (2004)
Electrical distribution and installation
Plant Engineer's Reference Book (2002)
C D Liapis et al.
Anticancer research, 25(3c), 2449-2452 (2005-08-06)
Carotid body (CB) is a highly specialized paraganglion originating from the neural crest ectoderm. CB paraganglion can be caused either by a genetic predisposition (hereditary paraganglia) or by chronic hypoxic stimulation. Germline mutations in any of the following genes: SDHD



Global Trade Item Number

SKUGTIN
M5904-5ML04061837898488
M5904-6X500ML04061837898501
M5904-5X5ML04061837898495
M5904-500ML04061837898471

Questions

1–4 of 4 Questions  
  1. What is the Specific Heat Capacity of this product?

    1 answer
    1. For catalog number M5904, we have not determined the heat capacity value. Here is the link for our SDS:
      https://www.sigmaaldrich.com/US/en/sds/sigma/m5904?userType=anonymous

      Helpful?

  2. Is this mineral oil suitable for microaerobic or anaerobic cultures of E. coli?

    1 answer
    1. This product has not been tested for cell culture use. However, in the literature, it is known to work with aerobic and anaerobic cell cultures.

      Helpful?

  3. Hello, I´m looking for a mineral oil for sulfur pattern production. The COA of this product says "pass" for sufurous compounds, what does this mean?

    1 answer
    1. The production and preparation of this product are considered proprietary. Currently, there is no product option available that is tested for the presence of sulfur.

      Helpful?

  4. Esiste una data di scadenza per il prodotto in questione?

    1 answer
    1. The product specification sheet for this product contains a recommended retest date period of roughly four years after the quality release date. A recommended retest date is the period of time during which the product is expected to remain within established stability specifications, provided that it has been stored under defined conditions. After the Retest Date, product samples should be examined to ensure that the product is still in compliance with the established specifications. Please view more information here: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/190/408/product-dating-information-ms.pdf.

      Helpful?

Reviews

No rating value

Active Filters