330760
Mineral oil
heavy
Synonym(s):
White mineral oil (petroleum)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Quality Level
form
liquid (colorless)
color
white
refractive index
n20/D 1.467 (lit.)
density
0.862 g/mL at 25 °C
InChI key
AEOVEGJBKQQFOP-DDVLFWKVSA-L
Looking for similar products? Visit Product Comparison Guide
General description
Mineral oil is made up of saturated and aromatic hydrocarbons. It is mainly found in hair oil formulations and liquid emollients used in cosmetics.
Application
Mineral oil has been used:
- in transplantation experiments
- in the preparation of Psuedomonas aeruginosa embedded beads
- for diluting benzaldehyde and 3-octanol, for its use in short-term memory experiments
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
No data available
Flash Point(C)
No data available
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Xenotransplantation of embryonic stem cell-derived motor neurons into the developing chick spinal cord
Stem Cells in Regenerative Medicine (2009)
Aromatic hydrocarbons of mineral oil origin in foods: a method for determining the total concentration and first results
Journal of Agricultural and Food Chemistry, 57(19), 8711-8721 (2009)
Study of sensory properties of emollients used in cosmetics and their correlation with physicochemical properties
International Journal of Cosmetic Science, 27(6), 354-354 (2005)
The involvement of potassium channel ORK1 in short-term memory and sleep in Drosophila
Medicine, 96(27) (2017)
A lung segmental model of chronic Pseudomonas infection in sheep.
PLoS ONE, 8(7) (2013)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service