Skip to Content
Merck

Skip To

I2149

Hemagglutinin Peptide

≥97% (HPLC), powder, Influenza

Synonym(s):

Anti-HA, HA tag peptide, HA1 fragment 98-106, Tyr-Pro-Tyr-Asp-Val-Pro-Asp-Tyr-Ala

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C53H67N9O17
CAS Number:
Molecular Weight:
1102.15
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.56
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Influenza Hemagglutinin (HA) Peptide, ≥97% (HPLC)

biological source

synthetic

Quality Level

assay

≥97% (HPLC)

form

powder

mol wt

1102.15 Da

technique(s)

protein extraction: suitable

storage temp.

−20°C

SMILES string

CC(C)[C@H](NC(=O)[C@H](CC(O)=O)NC(=O)[C@H](Cc1ccc(O)cc1)NC(=O)[C@@H]2CCCN2C(=O)[C@@H](N)Cc3ccc(O)cc3)C(=O)N4CCC[C@H]4C(=O)N[C@@H](CC(O)=O)C(=O)N[C@@H](Cc5ccc(O)cc5)C(=O)N[C@@H](C)C(O)=O

InChI

1S/C53H67N9O17/c1-27(2)44(52(77)62-21-5-7-41(62)50(75)59-38(25-42(66)67)47(72)56-36(45(70)55-28(3)53(78)79)23-30-10-16-33(64)17-11-30)60-48(73)39(26-43(68)69)57-46(71)37(24-31-12-18-34(65)19-13-31)58-49(74)40-6-4-20-61(40)51(76)35(54)22-29-8-14-32(63)15-9-29/h8-19,27-28,35-41,44,63-65H,4-7,20-26,54H2,1-3H3,(H,55,70)(H,56,72)(H,57,71)(H,58,74)(H,59,75)(H,60,73)(H,66,67)(H,68,69)(H,78,79)/t28-,35-,36-,37-,38-,39-,40-,41-,44-/m0/s1

InChI key

HVLSXIKZNLPZJJ-TXZCQADKSA-N

General description

Influenza Hemagglutinin (HA) peptide is a synthetic peptide with an amino acid sequence of N-Tyr-Pro-Tyr-Asp-Val-Pro-Asp-Tyr-Ala-C. This corresponds to amino acids 98-106 of the human influenza virus hemagglutinin. The HA Peptide is used for competing out the anti-HA antibody binding to HA-tagged fusion proteins in immunoassays. The successful inhibition of antibody binding by HA peptide demonstrates binding is specific.

Application

Influenza Hemagglutinin (HA) Peptide may be used in immunoblotting. It may also be used to to elute HA-tagged fusion proteins from an affinity column of Monoclonal Anti-HA agarose.

Preparation Note

Dissolve in water to a concentration of 5 mg/mL.

Other Notes

Lyophilized from 0.1% TFA in H2O

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
F3290P1306SAE0194
technique(s)

protein extraction: suitable

technique(s)

-

technique(s)

-

technique(s)

protein extraction: suitable

assay

≥97% (HPLC)

assay

≥85% (HPLC)

assay

≥97% (HPLC)

assay

≥95% (HPLC)

biological source

synthetic

biological source

-

biological source

synthetic

biological source

synthetic

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

form

powder

form

lyophilized powder

form

solid

form

lyophilized powder

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

−20°C


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Xiaolong Tang et al.
Nature communications, 8(1), 318-318 (2017-08-23)
Distant metastasis is the main cause of breast cancer-related death; however, effective therapeutic strategies targeting metastasis are still scarce. This is largely attributable to the spatiotemporal intratumor heterogeneity during metastasis. Here we show that protein deacetylase SIRT7 is significantly downregulated
Can Yang et al.
Developmental and comparative immunology, 100, 103426-103426 (2019-07-02)
Interferon regulated factor 5 (IRF5) is a key regulator of inflammatory responses in human and mammals; however, its role in teleost remains largely unknown. In this study, IRF5 homologue of black carp (Mylopharyngodon piceus) has been cloned and characterized, which
Sheng-Na Li et al.
Journal of medical virology, 91(9), 1668-1678 (2019-05-21)
Retinoic acid-inducible gene-I (RIG-I) belongs to the RIGI-like receptors (RLRs), a class of primary pattern recognition receptors. It senses viral double-strand RNA in the cytoplasm and delivers the activated signal to its adaptor virus-induced signaling adapter (VISA), which then recruits



Global Trade Item Number

SKUGTIN
I2149-.5MG04061833848319
I2149-1MG04061833848326

Questions

Reviews

No rating value

Active Filters