P1306
Peptide YY human
≥97% (HPLC), synthetic
Synonym(s):
PYY, Tyr-Pro-Ile-Lys-Pro-Glu-Ala-Pro-Gly-Glu-Asp-Ala-Ser-Pro-Glu-Glu-Leu-Asn-Arg-Tyr-Tyr-Ala-Ser-Leu-Arg-His-Tyr-Leu-Asn-Leu-Val-Thr-Arg-Gln-Arg-Tyr-NH2
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C194H295N55O57
CAS Number:
Molecular Weight:
4309.75
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.32
Recommended Products
Related Categories
Amino Acid Sequence
Tyr-Pro-Ile-Lys-Pro-Glu-Ala-Pro-Gly-Glu-Asp-Ala-Ser-Pro-Glu-Glu-Leu-Asn-Arg-Tyr-Tyr-Ala-Ser-Leu-Arg-His-Tyr-Leu-Asn-Leu-Val-Thr-Arg-Gln-Arg-Tyr-NH2
General description
Human peptide YY (PYY) is secreted by mucosal L cells in the distal ileum, colon and rectum. It consists of 36 amino acids. The gene encoding PYY is localized on human chromosome 17q21.1.
Application
Peptide YY human has been used as a supplement in Roswell Park Memorial Institute medium (RPMI medium) to culture pancreatic duct-derived cells.
Biochem/physiol Actions
Gut hormone that inhibits both secretin and cholecystokinin-stimulated pancreatic secretion. Endogenous nonselective agonist at NPY receptors.
Human peptide YY (PYY) has a role in food ingestion, gut motility and insulin secretion. The protein suppresses appetite. It has been associated with obesity and type 2 diabetes. PYY also has a role in energy homeostasis.
Inhibits secretin and cholecystokinin-stimulated pancreatic secretion;endogenous nonselective agonist at NPY receptors.
Other Notes
Lyophilized from 0.1% TFA in H2O
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
TLR ligands and butyrate increase Pyy expression through two distinct but inter-regulated pathways.
Larraufie P
Cellular Microbiology (2017)
Genomic and gene expression signature of the pre-invasive
testicular carcinoma in situ
testicular carcinoma in situ
Kristian Almstrup
Cell and Tissue Research (2005)
Rapid and Efficient In Vitro Generation of Pancreatic Islet
Progenitor Cells from Nonendocrine Epithelial Cells in the
Adult Human Pancreas
Progenitor Cells from Nonendocrine Epithelial Cells in the
Adult Human Pancreas
JOSEPHINE S. D?ALESSANDRO
Stem Cells and Development (2007)
Expression of peptide YY by human blood leukocytes.
Holler JP
Peptides (2014)
The Association of Serum Total Peptide YY (PYY) with Obesity and Body Fat Measures in the CODING Study.
Cahill F
PLoS ONE (2014)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service