Skip to Content
Merck

Skip To

F5259

Anti-Human IgA (α-chain specific)−FITC antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Goat Anti-Human IgA (α-chain specific)−Fluorescein isothiocyanate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46
MDL number:
Conjugate:
FITC conjugate
Clone:
polyclonal
Application:
IF (d)
Citations:
7
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

goat

Quality Level

conjugate

FITC conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

technique(s)

direct immunofluorescence: 1:16

storage temp.

2-8°C

target post-translational modification

unmodified

General description

IgA antibody is secretory antibody and is present abundantly in mucous linings of gastrointestinal, respiratory and genitourinary tracts, tears and saliva. Thus secretory IgA contributes to the humoral defense mechanism against the pathogens on mucosal surfaces
Anti-Human IgA (α-chain specific)-FITC antibody is specific for α chain of human IgA. Goat Anti-Human IgA is conjugated to Fluorescein Isothiocyanate (FITC), Isomer I.

Immunogen

Purified human IgA

Application

Anti-Human IgA (α-chain specific)-FITC antibody may be used for direct immunofluorescence at a working antibody dilution of 1:16 to 1:32. A dilution of 1:100 was used for immunohistochemistry of cryosections of human sural nerves.

Physical form

Solution in 0.01 M phosphate buffered saline pH 7.4, containing 1% BSA and 15 mM sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
F5384A6029A3187
biological source

goat

biological source

goat

biological source

goat

biological source

goat

technique(s)

direct immunofluorescence: 1:16

technique(s)

direct immunofluorescence: 1:32

technique(s)

direct ELISA: 1:10,000

technique(s)

direct ELISA: 1:30,000, immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50, western blot: 1:30,000

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

conjugate

FITC conjugate

conjugate

FITC conjugate

conjugate

peroxidase conjugate

conjugate

alkaline phosphatase conjugate

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

2-8°C

form

buffered aqueous solution

form

buffered aqueous solution

form

buffered aqueous solution

form

buffered aqueous glycerol solution


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content


G B Rosoklija et al.
Acta neuropathologica, 99(1), 55-62 (2000-01-29)
Quantitative immunocytochemical analysis of complement proteins (CP) was performed on sural nerve biopsies from 15 patients with diabetic neuropathy (DN) and 18 nondiabetic patients with other forms of chronic neuropathy (ON). The mean age of the patients and the pathological
Clayton A White et al.
Journal of immunology (Baltimore, Md. : 1950), 193(12), 5933-5950 (2014-11-14)
Class-switch DNA recombination (CSR) and somatic hypermutation (SHM), which require activation-induced cytidine deaminase (AID), and plasma cell differentiation, which requires B lymphocyte-induced maturation protein-1 (Blimp-1), are critical for the generation of class-switched and hypermutated (mature) Ab and autoantibody responses. We
Helia N Sanchez et al.
Nature communications, 11(1), 60-60 (2020-01-04)
Short-chain fatty acids (SCFAs) butyrate and propionate are metabolites from dietary fiber's fermentation by gut microbiota that can affect differentiation or functions of T cells, macrophages and dendritic cells. We show here that at low doses these SCFAs directly impact B



Global Trade Item Number

SKUGTIN
F5259-2ML04061833618141
F5259-1ML04061833618134

Questions

Reviews

No rating value

Active Filters