Skip to Content
Merck

Skip To

F5384

Anti-Human IgM (μ-chain specific)−FITC antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Goat Anti-Human IgM (μ-chain specific)−Fluorescein isothiocyanate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46
MDL number:
Conjugate:
FITC conjugate
Clone:
polyclonal
Application:
IF (d)
Citations:
13
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

goat

Quality Level

conjugate

FITC conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

technique(s)

direct immunofluorescence: 1:32

storage temp.

2-8°C

target post-translational modification

unmodified

General description

The product binds human IgM; but does not bind other human Igs.

Application

Anti-Human IgM (μ-chain specific)-FITC antibody produced in goat was used as secondary antibody in serodiagnosis of Lyme disease based on seroreactivity against Borrelia spirochaetes.

Biochem/physiol Actions

IgM antibodies are present as pentamers in the serum and are produced in response to antigens.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
F0132F4262A0420
conjugate

FITC conjugate

conjugate

FITC conjugate

conjugate

FITC conjugate

conjugate

peroxidase conjugate

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

IgG fraction of antiserum

antibody form

affinity isolated antibody

technique(s)

direct immunofluorescence: 1:32

technique(s)

direct immunofluorescence: 1:32

technique(s)

direct immunofluorescence: 1:32

technique(s)

direct ELISA: 1:50,000, dot blot: 1:100,000 (chemiluminescent), immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:200

biological source

goat

biological source

goat

biological source

goat

biological source

goat

form

buffered aqueous solution

form

buffered aqueous solution

form

buffered aqueous solution

form

-

clone

polyclonal

clone

polyclonal

clone

polyclonal

clone

polyclonal


Still not finding the right product?

or

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content


Nayoung Ko et al.
Scientific reports, 12(1), 9611-9611 (2022-06-11)
Pig-to-human organ transplantation is a feasible solution to resolve the shortage of organ donors for patients that wait for transplantation. To overcome immunological rejection, which is the main hurdle in pig-to-human xenotransplantation, various engineered transgenic pigs have been developed. Ablation
First detection and molecular identification of Borrelia garinii isolated from human skin in Taiwan.
Li-Lian Chao et al.
Journal of medical microbiology, 59(Pt 2), 254-257 (2009-11-17)
Jingtao Cui et al.
PloS one, 13(11), e0206995-e0206995 (2018-11-08)
Chlamydia pneumoniae (C. pneumoniae) is an obligate intracellular bacterium and a human pathogen that causes respiratory infectious diseases. More than 50% of the adult population worldwide was once infected with C. pneumoniae, but investigations into this topic are insufficient in



Global Trade Item Number

SKUGTIN
F5384-.5ML04061838354495
F5384-1ML04061837516894
F5384-2ML04061837516900

Questions

Reviews

No rating value

Active Filters