Skip to Content
Merck

Skip To

F1392

Riboflavin 5′-monophosphate sodium salt hydrate

suitable for electrophoresis, suitable for acrylamide photopolymerization, ≥70%, powder

Synonym(s):

FMN-Na, Flavin mononucleotide, Riboflavin 5′-phosphate sodium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C17H20N4NaO9P · xH2O
CAS Number:
Molecular Weight:
478.33 (anhydrous basis)
UNSPSC Code:
41106305
NACRES:
NA.51
PubChem Substance ID:
EC Number:
204-988-6
Beilstein/REAXYS Number:
4106529
MDL number:
eCl@ss:
34058005
Assay:
≥70%
Form:
powder
Solubility:
H2O: soluble 50 mg/mL, clear, orange
Storage temp.:
−20°C
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

assay

≥70%

form

powder

optical activity

[α]20/D +37 to +42°, c = 1.5 in 5 M HCl(lit.)

technique(s)

electrophoresis: suitable

solubility

H2O: soluble 50 mg/mL, clear, orange

ε (extinction coefficient)

12.5 at 445 nm at 1 mM

suitability

suitable for acrylamide photopolymerization

foreign activity

Protease, none detected

storage temp.

−20°C

SMILES string

O.[Na+].Cc1cc2N=C3C(=O)NC(=O)N=C3N(C[C@H](O)[C@H](O)[C@H](O)COP(O)([O-])=O)c2cc1C

InChI

1S/C17H21N4O9P.Na.H2O/c1-7-3-9-10(4-8(7)2)21(15-13(18-9)16(25)20-17(26)19-15)5-11(22)14(24)12(23)6-30-31(27,28)29;;/h3-4,11-12,14,22-24H,5-6H2,1-2H3,(H,20,25,26)(H2,27,28,29);;1H2/q;+1;/p-1/t11-,12+,14-;;/m0../s1

InChI key

BHRVCJBIICJWTH-APQIITSESA-M

Application

Riboflavin 5′-monophosphate sodium salt hydrate or FMN is suitable:
  • to study its mechanism of inhibition of mutagenicity of a benzo[a]pyrene 7,8-diol 9,10-epoxide, a carcinogenic metabolite of benzo[a]pyrene[1]
  • along with flavin adenine dinucleotide for reactivation of the enzyme that catalyzes the oxidation of pyridoxamine to pyridoxal in a study[2]
  • to study the reduction of polymeric azo and nitro dyes by intestinal bacteria[3]
  • to study the effect of FMN on ferrisiderophore reductase activity in the cytoplasmic fraction of Agrobacterium tumefaciens.[4]

Biochem/physiol Actions

Riboflavin 5′-monophosphate or Flavin mononucleotide (FMN) serves as a prosthetic group for flavoenzymes.[5] It is a part of the mitochondrial respiratory chain complex I and acts as a redox carrier. FMN is unstable in alkaline solution and is highly stable at pH 6.[6]
FMN is a coenzyme synthesized from riboflavin (vitamin B2). It acts as a component of complex I of the electron transport chain. The enzymes containing FMN are called as flavin enzymes and are involved in oxidation-reduction mechanisms.
FMN is suitable as a photopolymerization reagent in polyacrylamide gel electrophoresis (PAGE) by forming free radicals in aqueous solution in the presence of light. FMN photodecomposes to leucoflavin. No free radicals are formed in the absence of oxygen, but traces of oxygen allow for leucoflavin to reoxidize with free-radical generation. The catalysts, tetramethylethylenediamine (TEMED) or 3-dimethylaminopropionitrile (DMAPN), are commonly added to speed up the free radical formation. Free radicals will cause acrylamide and bis-acrylamide to polymerize to form a gel matrix which can be used for sieving macromolecules. FMN is commonly used in the stacking gel for non-denaturing PAGE because native proteins can be sensitive to persulfate ions from ammonium persulfate. Another advantage of FMN over ammonium persulfate is that it will not start polymerizing until the gel is illuminated.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
F2253F675077623
Quality Level

100

Quality Level

200

Quality Level

200

Quality Level

200

form

powder

form

powder

form

powder

form

powder

assay

≥70%

assay

≥70% (HPLC)

assay

70-79% (fluorimetric)

assay

-

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

2-8°C

solubility

H2O: soluble 50 mg/mL, clear, orange

solubility

water: 50 mg/mL

solubility

water: 50 mg/mL, orange

solubility

water: soluble 100 g/L at 20 °C

technique(s)

electrophoresis: suitable

technique(s)

-

technique(s)

-

technique(s)

-


Still not finding the right product?


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



J S Lodge et al.
Journal of bacteriology, 149(2), 771-774 (1982-02-01)
Reduction of the iron in ferriagrobactin by the cytoplasmic fraction of Agrobacterium tumefaciens strictly required NaDH as the reductant. Addition of flavin mononucleotide and anaerobic conditions were necessary for the reaction; when added with flavin mononucleotide, magnesium was stimulatory. This
A W Wood et al.
Proceedings of the National Academy of Sciences of the United States of America, 79(17), 5122-5126 (1982-09-01)
Riboflavin 5'-phosphate (flavin mononucleotide; FMN) inhibits the mutagenicity of (+/-)-7 beta, 8 alpha-dihydroxy-9 alpha, 10 alpha-epoxy-7,8,9,10-tetrahydrobenzo[a]pyrene (B[a]P diol epoxide), the only known ultimate carcinogenic metabolite of benzo[a]pyrene. Coincubation of 10, 25, and 50 nmol of FMN with strain TA100 of
Matthias Mack et al.
Applied microbiology and biotechnology, 71(3), 265-275 (2006-04-12)
Flavins are active components of many enzymes. In most cases, riboflavin (vitamin B(2)) as a coenzyme represents the catalytic part of the holoenzyme. Riboflavin is an amphiphatic molecule and allows a large variety of different interactions with the enzyme itself



Global Trade Item Number

SKUGTIN
F1392-25MG04061833611937

Questions

Reviews

No rating value

Active Filters