Skip to Content
Merck

F6750

Riboflavin 5′-monophosphate sodium salt hydrate

70-79% (fluorimetric)

Synonym(s):

FMN-Na, Flavin mononucleotide, Riboflavin 5′-phosphate sodium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C17H20N4NaO9P · xH2O
CAS Number:
Molecular Weight:
478.33 (anhydrous basis)
NACRES:
NA.51
PubChem Substance ID:
eCl@ss:
34058005
UNSPSC Code:
41106305
MDL number:
Beilstein/REAXYS Number:
4106529
Assay:
70-79% (fluorimetric)
Biological source:
non-animal source
Form:
powder
Solubility:
water: 50 mg/mL, orange
Storage temp.:
−20°C
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

non-animal source

Quality Segment

description

(Clear to hazy)

assay

70-79% (fluorimetric)

form

powder

solubility

water: 50 mg/mL, orange

storage temp.

−20°C

SMILES string

O.[Na+].Cc1cc2N=C3C(=O)NC(=O)N=C3N(C[C@H](O)[C@H](O)[C@H](O)COP(O)([O-])=O)c2cc1C

InChI

1S/C17H21N4O9P.Na.H2O/c1-7-3-9-10(4-8(7)2)21(15-13(18-9)16(25)20-17(26)19-15)5-11(22)14(24)12(23)6-30-31(27,28)29;;/h3-4,11-12,14,22-24H,5-6H2,1-2H3,(H,20,25,26)(H2,27,28,29);;1H2/q;+1;/p-1/t11-,12+,14-;;/m0../s1

InChI key

BHRVCJBIICJWTH-APQIITSESA-M

General description

Riboflavin 5′-monophosphate is a derivative of riboflavin, a water-soluble vitamin.

Application

Riboflavin 5′-monophosphate sodium salt hydrate has been used in the preparation of hypo-osmolar riboflavin formulation:
  • to evaluate the riboflavin (RF) concentration and distribution in the corneal stroma
  • to study corneal resistance to enzymatic digestion
  • as a photosensitizer to evaluate the efficacy and safety of photochemical corneal stiffening using ex vivo and in vivo rabbit eye models

Riboflavin 5′-monophosphate is used as a substrate to study the specificity and kinetics of FMN phosphohydrolases. FMN is used to study molecular sensing based gene regulation by riboswitches.

Biochem/physiol Actions

Riboflavin 5′-monophosphate (FMN) serves as a cofactor for flavoenzymes. It is used as a prosthetic group by oxidoreductase (one and two-electron transfer) enzymes, also called flavoproteins, such as nicotinamide adenine dinucleotide (NADH) dehydrogenases, nitric oxide synthases, nitric oxide reductase, and nitrilotriacetate monooxygenases. These enzymes protect cells from oxidative stress and apoptosis. Any alteration in the levels of FMN caused due to abnormal riboflavin metabolism leads to mitochondrial dysfunction and diminished energy levels. This results in disorders related to riboflavin such as cataracts, neurodegenerative and cardiovascular diseases. FMN is associated with metabolic pathways of amino acids, fatty acids, and purines. It is involved in cellular growth and development mediated by the oxidation-reduction reaction.

Analysis Note

Free Riboflavin: ≤6.0%
Riboflavin diphosphates: ≤6.0% (as riboflavin)


Still not finding the right product?


Storage Class

11 - Combustible Solids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library