Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
NACRES:
NA.55
UNSPSC Code:
12352200
Pricing and availability is not currently available.
1 of 1
This Item | |||
|---|---|---|---|
| color colorless | color - | color - | color - |
| shipped in wet ice | shipped in - | shipped in - | shipped in wet ice |
| Quality Level 200 | Quality Level 200 | Quality Level 200 | Quality Level 200 |
| grade Molecular Biology | grade - | grade Molecular Biology | grade - |
| storage temp. −20°C | storage temp. −20°C | storage temp. room temp | storage temp. −20°C |
| feature hotstart | feature Direct PCR | feature - | feature Direct PCR |
Storage Class
12 - Non Combustible Liquids
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Holly V Moeller et al.
Ecology and evolution, 6(15), 5416-5430 (2016-08-24)
Theory predicts that neighboring communities can shape one another's composition and function, for example, through the exchange of member species. However, empirical tests of the directionality and strength of these effects are rare. We determined the effects of neighboring communities
Competition-function tradeoffs in ectomycorrhizal fungi.
Moeller HV and Peay KG
PeerJ, 4, e2270-e2270 (2016)
Daniela Hegebarth et al.
Plant, cell & environment, 40(9), 1761-1776 (2017-05-10)
The aliphatic waxes sealing plant surfaces against environmental stress are generated by fatty acid elongase complexes, each containing a β-ketoacyl-CoA synthase (KCS) enzyme that catalyses a crucial condensation forming a new C─C bond to extend the carbon backbone. The relatively
Global Trade Item Number
| SKU | GTIN |
|---|---|
| E7526-240ML | 04061838248572 |
| E7526-12ML | 04061838248565 |
| E7526-1L | 04061833608401 |
| E7526-24ML | 04061833608418 |
| E7526-60ML | 04061838248602 |
| E7526-1.2ML | 04061838247599 |
| E7526-120ML | 04061833605172 |



