Skip to Content
Merck

Key Documents

Skip To

T3073

Tissue Preparation Solution

30 mL sufficient for 1,000 extractions

Synonym(s):

Direct PCR tissue preparation solution, Extract-N-Amp™ Tissue Preparation Solution, REDExtract-N-Amp™ Tissue Preparation Solution, SYBR™ Green Extract-N-Amp™ Tissue Preparation Solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352200
EC Number:
254-457-8
NACRES:
NA.55
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

usage

30 mL sufficient for 1,000 extractions, sufficient for 1000 reactions

feature

Direct PCR

technique(s)

PCR: suitable

input

crude DNA

shipped in

wet ice

storage temp.

−20°C

General description

The Tissue Preparation Solution is intended for use with the Extract-N-Amp™ Tissue PCR kits, REDExtract-N-Amp™ Tissue PCR kits, and SYBR™ Green Extract-N-Amp Tissue PCR kits. This product is intended to supplement the kit reagents, where there are many extractions.

Application

Tissue preparation solution has been used:
  • in combining segments of tails for genotyping
  • to extract genomic DNA from zebrafish embryos/fin tissues
  • to extract DNA from ear tags, tail tips and embryonic yolk sacs of mouse embryo
  • for extraction of DNA from fungal spores and mycelia

Legal Information

SYBR is a trademark of Thermo Fisher Scientific or its subsidiaries
Extract-N-Amp is a trademark of Sigma-Aldrich Co. LLC
REDExtract-N-Amp is a trademark of Sigma-Aldrich Co. LLC

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
XNATRGXNATGXNAB
description

30 mL sufficient for 1,000 extractions

description

sufficient for 1000 extractions, sufficient for 1000 amplifications

description

sufficient for 100 extractions, sufficient for 100 amplifications

description

sufficient for 100 extractions, sufficient for 100 amplifications

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

input

crude DNA

input

crude DNA

input

crude DNA

input

crude DNA

technique(s)

PCR: suitable

technique(s)

PCR: suitable, whole genome amplification: suitable

technique(s)

PCR: suitable, whole genome amplification: suitable

technique(s)

whole genome amplification: suitable, PCR: suitable

shipped in

wet ice

shipped in

-

shipped in

-

shipped in

wet ice

usage

sufficient for 1000 reactions, 30 mL sufficient for 1,000 extractions

usage

sufficient for 1000 reactions, sufficient for 1000 extractions, sufficient for 1000 amplifications

usage

sufficient for 100 extractions, sufficient for 100 reactions, sufficient for 100 amplifications

usage

sufficient for 100 extractions, sufficient for 100 reactions, sufficient for 100 amplifications

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class

10 - Combustible liquids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

Active Filters

  1. 1 Ratings-only review