Skip to Content
Merck
All Photos(3)

Documents

E5750

Sigma-Aldrich

Ergocalciferol

40,000,000 USP units/g

Synonym(s):

Calciferol, Ercalciol, Ergosterol irradiated, Irradiated ergosterol, Vitamin D2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H44O
CAS Number:
Molecular Weight:
396.65
Beilstein:
1916682
EC Number:
MDL number:
UNSPSC Code:
12352205
eCl@ss:
34058017
PubChem Substance ID:
NACRES:
NA.79

biological source

synthetic (organic)

Quality Level

Assay

≥97%

form

powder

specific activity

40,000,000 USP units/g

technique(s)

HPLC: suitable

color

white to off-white

mp

114-118 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(C)[C@@H](C)/C=C/[C@@H](C)[C@@]1([H])CC[C@@]([C@]1(C)CCC/2)([H])C2=C\C=C(C[C@@H](O)CC3)/C3=C

InChI

1S/C28H44O/c1-19(2)20(3)9-10-22(5)26-15-16-27-23(8-7-17-28(26,27)6)12-13-24-18-25(29)14-11-21(24)4/h9-10,12-13,19-20,22,25-27,29H,4,7-8,11,14-18H2,1-3,5-6H3/b10-9+,23-12+,24-13-/t20-,22+,25-,26+,27-,28+/m0/s1

InChI key

MECHNRXZTMCUDQ-RKHKHRCZSA-N

Gene Information

human ... VDR(7421)

Looking for similar products? Visit Product Comparison Guide

General description

Vitamin D2 or ergocalciferol is available through dietary supplements or plant sources. It is a provitamin, which is fat-soluble.

Application

Ergocalciferol has been used as a component of the diet for rats, to study its bioavailability from ultraviolet-irradiated D2-rich yeast. It has also been used as a standard for identification and quantification of vitamins, as part of a study to determine the nutritional composition of food fishes.

Biochem/physiol Actions

Ergocalciferol (vitamin D2) and 25-Hydroxycholecalciferol (vitamin D3) are the two forms of vitamin D, which are activated in vivo by hydroxylation. Vitamin D2 and D3 may be used in a wide range of studies to assess their effects on function such as immune function and calcium homeostasis.
Ergocalciferol is used for antirachitic treatment. Vitamin D has an active role in bone mineralization. Severe deficiency of this vitamin has been associated with osteomalacia and rickets.

Packaging

Sealed ampule.

Quantity

1 USP unit = 1 I.U.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - STOT RE 1 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Poor bioavailability of vitamin D2 from ultraviolet-irradiated D2-rich yeast in rats
Itkonen ST, et al.
Nutrition Research (New York, N.Y.), 59, 36-43 (2018)
Hypoparathyroidism and Other Causes of Hypocalcemia
Luciano M
Encyclopedia of Endocrine Diseases (2018)
Nutritional composition of food fishes and their importance in providing food and nutritional security
Mohanty BP, et al.
Food Chemistry, 293 (2019)
Dietary Reference Intakes for Calcium and Vitamin D (2011)
Vitamin D: The Biotechnology of Ergosterol
Vandamme EJ
Biotechnology of microbial enzymes (1989)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service