Skip to Content
Merck

Skip To

42579

4-Phenyl-1,2,4-triazoline-3,5-dione

derivatization grade (HPLC), LiChropur, ≥98.0% (CHN)

Synonym(s):

Cookson reagent, 4-Phenyl-3H-1,2,4-triazole-3,5(4H)-dione, PTAD

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C8H5N3O2
CAS Number:
Molecular Weight:
175.14
UNSPSC Code:
41116105
NACRES:
NA.22
PubChem Substance ID:
EC Number:
224-191-7
Beilstein/REAXYS Number:
141548
MDL number:
Assay:
≥98.0% (CHN)
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

derivatization grade (HPLC)

Quality Level

assay

≥98.0% (CHN)

quality

LiChropur

technique(s)

HPLC: suitable

mp

165-170 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

O=C1N=NC(=O)N1c2ccccc2

InChI

1S/C8H5N3O2/c12-7-9-10-8(13)11(7)6-4-2-1-3-5-6/h1-5H

InChI key

ISULLEUFOQSBGY-UHFFFAOYSA-N

General description

4-Phenyl-1,2,4-triazoline-3,5-dione, also known as Cookson reagent, is a strong dienophile, which gives a stable Diels-Alder adduct quantitatively within a short time and under mild conditions. It is commonly used as a protecting group of the diene moiety for the synthesis of vitamin D3 (VD3)-related compounds.

Application

4-Phenyl-1,2,4-triazoline-3,5-dione may be used as a derivatizing reagent for the determination of trace levels of 25-hydroxyvitamin D and its C-3 epimer in biological samples and cholecalciferol (vitamin D3) in fortified infant formula, milk and milk powder[1] using liquid chromatography–tandem mass spectrometry (LC–MS/MS)technique.

Other Notes

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Chromatographic separation of PTAD-derivatized 25-hydroxyvitamin D3 and its C-3 epimer from human serum and murine skin
Teegarden DM, et al.
Journal of Chromatography. B, Analytical Technologies in the Biomedical and Life Sciences, 991(1), 118-121 (2015)
Dienophilic Reagent for Precolumn Derivatization of 7-Dehydrocholesterol in High Performance Liquid Chromatography
Shimada K and OE T
Analytical Sciences, 6(3), 461-463 (1990)
A S Weiskopf et al.
Journal of mass spectrometry : JMS, 36(1), 71-78 (2001-02-17)
The structural specificity of vitamin D derivatization by PTAD (4-phenyl-1,2,4-triazoline-3,5-dione) was probed using synthetic analogues and ion trap mass spectrometry. EB 1089, a vitamin D(3) analogue which contains a second site for Diels--Alder cycloaddition on its side-chain, allowed the examination



Global Trade Item Number

SKUGTIN
42579-10X100MG04061833439135
42579-100MG04061832104065

Questions

Reviews

No rating value

Active Filters