Skip to Content
Merck

Skip To

D8899

Deoxyribonucleic acid, single stranded from calf thymus

lyophilized powder

Synonym(s):

DNA single stranded

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
41106310
eCl@ss:
32160414
EC Number:
293-507-3
NACRES:
NA.52
MDL number:
Biological source:
bovine (calf) thymus
Form:
lyophilized powder
Storage temp.:
−20°C
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

bovine (calf) thymus

Quality Level

grade

Molecular Biology

form

lyophilized powder

mol wt

~50 kb

composition

ssDNA, ≥65%

storage temp.

−20°C

General description

High quality single-stranded template DNA isolated from the thymus of male and female calves.

Application

Calf thymus DNA is exceptionally useful as a substrate for DNA polymerase assays, in the amplification of very long fragments and as a carrier DNA for precipitations.
Calf thymus DNA is exceptionally useful as a substrate for DNA polymerase assays, in the amplification of very long fragments and as a carrier DNA for precipitations. It was used as a standard for DNA quantification by fluorescent assay[1] and in DNA-binding assay.[2]

Features and Benefits

• Activated
• No nuclease activity detected after 16 h at 37 °C incubation
• High quality template DNA

Preparation Note

Dissolve the DNA gently rocking overnight to a final concentration of 1mg/ml in molecular biology grade water.
One mg DNA is approx. 25 A260 units.
Prepared by a modification of the method of Alberts and Herrick from calf thymus DNA.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
D4522D4764D8273
grade

Molecular Biology

grade

Molecular Biology

grade

Molecular Biology

grade

-

biological source

bovine (calf) thymus

biological source

bovine pancreas

biological source

bovine (calf) thymus

biological source

bovine (calf) thymus

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

storage temp.

−20°C

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

−20°C

mol wt

~50 kb

mol wt

-

mol wt

>20 kb

mol wt

-


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content

Product Information Sheet


H Kamei
Cell biology international, 20(11), 769-775 (1996-11-01)
The anti-desmin monoclonal antibodies (MAbs) DSB389, AC54, and DSB860 recognize intermediate filaments (IFs) and nuclear antigens that appear granular, locate around chromosomes, and are insoluble following 0.5% Triton X-100 and cetyltrimethylammonium bromide (CTAB) extraction. Nuclear antigens of the MAbs were
J A Hansen et al.
Journal of animal science, 75(7), 1810-1821 (1997-07-01)
We evaluated the effects of recombinant porcine somatotropin (pST) and the beta-adrenergic agonist salbutamol on plasam endocrine and metabolite profiles and muscle chemistry in three genotypes of growing barrows (n = 96, 139 d old). Treatments were in a 2
Qiong Guo et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 106, 155-162 (2013-02-05)
A divanadium(V) complex, [V2O3(o-van-val)2] (o-van-val=Schiff base derived from o-vanillin and L-valine), has been synthesized and structurally characterized. The crystal structure shows that both of the vanadium centers in the complex have a distorted octahedral coordination environment composed of tridentate Schiff



Global Trade Item Number

SKUGTIN
D8899-1MG04061833590065
D8899-5MG04061833289525

Questions

  1. Does the product reanneal after storage and cooling at room temperature? Should we boil the ssDNA prior to use? How many base pairs are included in this ssDNA?

    1 answer
    1. Unfortunately, information regarding the reanneal after storing and cooling at room temperature has not been determined.

      Product D8899 is listed as having a length of approximately 50 kb. For single stranded DNA, the molecular weight of one kb is approximately equal to 3.3 × 105 daltons, so 50 kb has a weight of approximately 16.5 × 106 daltons.

      Helpful?

Reviews

No rating value

Active Filters