Skip to Content
Merck
All Photos(1)

Key Documents

D4764

Sigma-Aldrich

Deoxyribonucleic acid from calf thymus

Genomic, unsheared

Synonym(s):

double-stranded template DNA, Calf Thymus DNA, DNA from calf thymus, Thymonucleic acid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
41106310
eCl@ss:
32160414
NACRES:
NA.52
Pricing and availability is not currently available.

grade

Molecular Biology
for molecular biology

form

lyophilized powder

mol wt

>20 kb

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

High quality double-stranded template DNA isolated from the thymus of male and female calves.

Specificity

42% GC; Tm = 87 °C.
% GC is the percentage of G/C base pairs. Tm is the temperature at the midpoint of the thermal denaturation profile.

Application

Calf thymus DNA is exceptionally useful as a substrate for DNA polymerase assays, in the amplification of very long fragments and as a carrier DNA for precipitations.
Calf thymus DNA is exceptionally useful as a substrate for DNA polymerase assays, in the amplification of very long fragments and as a carrier DNA for precipitations. It was used as a standard for DNA quantification by fluorescent assay.[1] It was used as reference to determine the DNA content in cartilage tissue digest samples.[2][3]

Features and Benefits

• No nuclease activity detected after 16 h at 37 °C incubation
• High quality template DNA

Preparation Note

Purified by anion exchange chromatography, dialyzed against a solution of 1 mM sodium chloride, 1 mM EDTA and 1 mM Trizma HCl, pH 7.5 and lyophilized at a concentration of ~2 A260 units per mL. ~20 A260 units per mg DNA.

Reconstitution

Dissolve the DNA gently rocking overnight to a final concentration of 1mg/ml in molecular biology grade water.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Châtel et al.
Ecotoxicology (London, England), 20(8), 1727-1740 (2011-06-10)
In the present study, we analyzed the effects of two major pollutants of the environment, tributyltin (TBT) and water-accommodated fraction (WAF) of diesel oil, on MAP kinase activation, apoptosis induction and DNA damage, in the marine sponge Suberites domuncula. Our
Roel J H Custers et al.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society, 28(4), 429-435 (2009-11-04)
The purposes of the present study were to explore the surgical possibilities for replacement of the medial tibial plateau by a metallic implant in a large animal model and to examine the implications for the opposing cartilage. In six goats
Nathalie W D Jansen et al.
Arthritis and rheumatism, 56(1), 199-207 (2006-12-30)
Joint bleeding, or hemarthrosis, leads in time to severe joint damage. This study was carried out to test the in vitro thresholds of exposure time and concentration that lead to irreversible joint damage, to add to the discussion on the
José Luis García-Giménez et al.
Journal of inorganic biochemistry, 121, 167-178 (2013-02-07)
Ternary copper(II) complexes [Cu(NST)2(phen)] (1) and [Cu(NST)2(NH3)2]·H2O (2) [HNST=N-(4,5-dimethylthiazol-2-yl)naphthalene-1-sulfonamide] were prepared and characterized by physico-chemical techniques. Both 1 and 2 were structurally characterized by X-ray crystallography. The crystal structures show the presence of a distorted square planar CuN4 geometry in
K Bishayee et al.
Cell proliferation, 46(2), 153-163 (2013-03-21)
Small aromatic compounds like flavonoids can intercalate with DNA molecules bringing about conformational changes leading to reduced replication and transcription. Here, we have examined one dietary flavonoid, quercetin (found in many fruit and vegetables), for possible anti-cancer effects, on HeLa

Questions

  1. how much is 1 unit (i.e. in ug or mg)?

    1 answer
    1. One unit, based on the A260 value, is approximately 50 µg. The general conversion formula is 20 units per milligram. The 0.25 UN package size will contain approximately 12.5 µg, the 1 jUN package size will contain approximately 50 µg, and the 5UN package size will contain approximately 250 µg.

      For additional product information, please see the link below to review the product datasheet:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/289/542/d4764dat.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service