Skip to Content
Merck
All Photos(1)

Documents

D4416

Sigma-Aldrich

Deoxyribonucleic acid, High molecular weight from BALB/c mouse, adult

solution

Synonym(s):

Deoxyribonucleic acid, high molecular weight

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41106310
NACRES:
NA.52

grade

for molecular biology

Quality Level

form

solution

mol wt

>50 kbp

shipped in

wet ice

storage temp.

2-8°C

General description

Highly purified genomic DNA isolated from adult BALB/c mice.

Application

Deoxyribonucleic acid, High molecular weight from BALB/c mouse, adult, is used as a target for amplification by polymerase chain reaction (PCR), restriction endonuclease digestion and as a carrier DNA for precipitations. It may also be used in library construction in bacteriophage lambda vectors and in Southern hybridization. It was used as a standard for DNA quantification by spectrophotometry and as a positive control in TaqMan PCR reactions.
Applications include use as a target for amplification by polymerase chain reaction (PCR), restriction endonuclease digestion and as a carrier DNA for precipitations. May also be used in library construction in bacteriophage lambda vectors and in Southern hybridization.

Components

The DNA is in a solution of 10mM Tris-HCl (pH 7.5) with 0.1mM EDTA.

Other Notes

Highly purified genomic DNA.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eleanor R Gray et al.
PloS one, 6(3), e18096-e18096 (2011-03-31)
Several studies have implicated a recently discovered gammaretrovirus, XMRV (Xenotropic murine leukaemia virus-related virus), in chronic fatigue syndrome and prostate cancer, though whether as causative agent or opportunistic infection is unclear. It has also been suggested that the virus can
Stéphane Hué et al.
Retrovirology, 7(1), 111-111 (2010-12-22)
Xenotropic murine leukaemia viruses (MLV-X) are endogenous gammaretroviruses that infect cells from many species, including humans. Xenotropic murine leukaemia virus-related virus (XMRV) is a retrovirus that has been the subject of intense debate since its detection in samples from humans
Per Levéen et al.
Blood, 106(13), 4234-4240 (2005-09-01)
We have taken advantage of the Cre/lox system to generate a mouse model with inducible deficiency of transforming growth factor beta receptor II (TbetaRII). Using this approach, transforming growth factor beta (TGF-beta) signaling deficiency can be restricted to the hematopoietic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service