Skip to Content
Merck

Skip To

D4889

Deoxyribonucleic acid sodium salt from Escherichia coli strain B

Genomic, unsheared

Synonym(s):

DNA

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
41106310
eCl@ss:
32160414
EC Number:
297-226-7
NACRES:
NA.52
MDL number:
Biological source:
Escherichia coli
Form:
lyophilized powder
Solubility:
water: ~1 g/L
Storage temp.:
2-8°C
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

Escherichia coli

Quality Level

grade

Molecular Biology

form

lyophilized powder

mol wt

>45 kb

solubility

water: ~1 g/L

storage temp.

2-8°C

General description

Escherichia coli strain B is most widely studied DNA after k-12. The genomes of E coli strain B and k-12 show differences in the IS sequence elements. Strain B have B-islands and express O-specific side-chain LPS. The product is unsheared genomic DNA.

Application

The DNAs are particularly useful for genomic analysis, including PCR, library construction in bacteriophage λ, and in heterologous pre-hybridization and hybridization protocols.
Deoxyribonucleic acid sodium salt from Escherichia coli strain B has been used for:
  • PCR reactions of genomic DNA of Gardnerella vaginalis[1]
  • nick translation and radiolabeling of dNTP[2]
  • generating standard curve for 16S rDNA quantification[3]
  • transfecting neutrophils[4]
  • testing the extraction methods[5]

Biochem/physiol Actions

50% GC; Tm = 90.5 °C

Preparation Note

Purified by anion exchange chromatography, dialyzed against a solution of 1 mM sodium chloride, 1 mM EDTA and 1 mM Trizma HCl, pH 7.5 and lyophilized at a concentration of approx. 2 units per mL. Approx. 20 A260 units per mg DNA. % GC is the percentage of G/C base pairs. Tm is the temperature at the midpoint of the thermal denaturation profile.

Analysis Note

Analyzed by gel electrophoresis and visualization after ethidium bromide staining, with single predominant band of 45 kB or greater.

Other Notes

DNA in solution will reanneal on standing at room temperature so it is recommended to boil the solution for 10 minutes and then cool on ice for at least 5 minutes prior to use.
One unit will yield an A260 of 1.0 in 1.0 ml of 1 mM Tris-HCl, pH 7.5, with 1 mM NaCl and 1 mM EDTA (1 cm light path).

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
D2001S3917D9768
grade

Molecular Biology

grade

-

grade

Molecular Biology

grade

Molecular Biology

biological source

Escherichia coli

biological source

-

biological source

-

biological source

-

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

form

lyophilized powder

form

fibers

form

liquid

form

lyophilized powder

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

2-8°C

solubility

water: ~1 g/L

solubility

water: 0.98-1.02 mg/mL, faintly hazy to hazy, colorless to light yellow

solubility

-

solubility

-


Still not finding the right product?


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



M Agarwal et al.
BJU international, 88(9), 868-870 (2002-02-20)
To investigate the possible role of Gardnerella vaginalis in interstitial cystitis (IC), using molecular methods to avoid difficulties with the culture and recovery of viable organisms, and the problems associated with the recovery of low numbers of culturable organisms. Thirty-three
Yue Huo et al.
Current microbiology, 76(1), 22-28 (2018-11-02)
A Gram-positive bacterium (DCY118T) was isolated from ginseng-cultivated soil in Gochang-gun, Republic of Korea. This isolate was assigned to the genus Ornithinimicrobium and is closely related to Ornithinimicrobium kibberense K22-20T (98.8%), O. pekingense DSM 21552T (98.5%), O. algicola JC311T (98.2%)
Joseph W Duris et al.
Water research, 47(1), 300-314 (2012-11-15)
The occurrence and distribution of fecal indicator bacteria (FIB) and bacterial and protozoan pathogens are controlled by diverse factors. To investigate these factors in Pennsylvania streams, 217 samples were collected quarterly from a 27-station water-quality monitoring network from July 2007



Global Trade Item Number

SKUGTIN
D4889-.25UN04061833492628
D4889-1UN04061833492635
D4889-5UN04061833492642

Questions

1–2 of 2 Questions  
  1. D4889 Sigma-Aldrich Deoxyribonucleic acid sodium salt from Escherichia coli strain B I have 1 Unit of this DNA. Just want to make sure if it =50ug. Also, Can i use TE buffer to dilute the DNA? thank you for your help.

    1 answer
    1. One unit of DNA equals 50 µg. Molecular biology grade TE buffer is suitable for diluting DNA.

      Helpful?

  2. what is the absolute amount of DNA in this product is it 50µg?

    1 answer
    1. This product is supplied in a unit size. 1 unit of any DNA = 50 µg.

      Helpful?

Reviews

No rating value

Active Filters