Skip to Content
Merck

Skip To

D0629

3,5-Diiodo-L-thyronine

thyroid hormone analog

Synonym(s):

3,5-Diiodo-4-(4-hydroxyphenoxy)-L-phenylalanine, 3,5-T2, O-(4-Hydroxyphenyl)-3,5-diiodo-L-tyrosine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C15H13I2NO4
CAS Number:
Molecular Weight:
525.08
UNSPSC Code:
41116107
NACRES:
NA.26
PubChem Substance ID:
EC Number:
213-867-7
Beilstein/REAXYS Number:
2707891
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

assay

≥99% (HPLC)

form

powder

technique(s)

ligand binding assay: suitable

color

white to off-white

mp

255 °C

storage temp.

−20°C

SMILES string

N[C@@H](Cc1cc(I)c(Oc2ccc(O)cc2)c(I)c1)C(O)=O

InChI

1S/C15H13I2NO4/c16-11-5-8(7-13(18)15(20)21)6-12(17)14(11)22-10-3-1-9(19)2-4-10/h1-6,13,19H,7,18H2,(H,20,21)/t13-/m0/s1

InChI key

ZHSOTLOTTDYIIK-ZDUSSCGKSA-N

Biochem/physiol Actions

3,5-Diiodo-L-thyronine ((T(2)) is an iodinated thyronine hormone that regulates gene activity affecting processes such as homeostasis, lipid metabolism and insulin resistance.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
719536D0754T2877
form

powder

form

powder

form

crystalline

form

powder

technique(s)

ligand binding assay: suitable

technique(s)

mass spectrometry (MS): suitable

technique(s)

ligand binding assay: suitable

technique(s)

transfection: suitable

assay

≥99% (HPLC)

assay

98% (CP)

assay

≥98.0% (HPLC)

assay

≥95% (HPLC)

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

mp

255 °C

mp

-

mp

200 °C (dec.) (lit.)

mp

234-238 °C (lit.)


Still not finding the right product?

Explore all of our products under 3,5-Diiodo-L-thyronine


hcodes

Hazard Classifications

Aquatic Chronic 3

Storage Class

11 - Combustible Solids

wgk

WGK 3

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content


Maria Moreno et al.
Thyroid : official journal of the American Thyroid Association, 18(2), 239-253 (2008-02-19)
The processes and pathways mediating the intermediary metabolism of carbohydrates, lipids, and proteins are all affected by thyroid hormones (THs) in almost all tissues. Particular attention has been devoted by scientists to the effects of THs on lipid metabolism. Among
Roberto Mangiullo et al.
Biochimica et biophysica acta, 1797(2), 233-240 (2009-11-03)
Besides triiodothyronine (T3), 3,5-diiodo-L-thyronine (T2) has been reported to affect mitochondrial bioenergetic parameters. T2 effects have been considered as independent of protein synthesis. Here, we investigated the effect of in vivo chronic T2 administration to hypothyroid rats on liver mitochondrial
Alessio Rochira et al.
Biochimica et biophysica acta, 1831(12), 1679-1689 (2013-08-21)
Thyroid hormone 3,5,3'-triiodo-l-thyronine (T3) is known to affect cell metabolism through both the genomic and non-genomic actions. Recently, we demonstrated in HepG2 cells that T3 controls the expression of SREBP-1, a transcription factor involved in the regulation of lipogenic genes.



Global Trade Item Number

SKUGTIN
D0629-5G04061832416076
D0629-500MG04061833557921

Questions

Reviews

No rating value

Active Filters