Select a Size
| Pack Size | SKU | Availability | Price |
|---|
About This Item
biological source
bovine
Quality Level
assay
≥98% (agarose gel electrophoresis)
form
lyophilized powder
purified by
heat shock fractionation
packaging
poly bottle of
origin
New Zealand origin
technique(s)
flow cytometry: suitable
pH
7, ~7 (1% in 0.15 M NaCl)
solubility
water: slightly soluble
UniProt accession no.
storage temp.
2-8°C
Gene Information
bovine ... ALB(280717)
General description
Application
Biochem/physiol Actions
Preparation Note
1 of 1
This Item | |||
|---|---|---|---|
| origin New Zealand origin | origin USA origin | origin USA origin | origin New Zealand origin |
| technique(s) flow cytometry: suitable | technique(s) ELISA: suitable, blood typing: suitable, microbiological culture: suitable | technique(s) immunohistochemistry: suitable | technique(s) western blot: suitable |
| biological source bovine | biological source bovine | biological source bovine | biological source bovine |
| assay ≥98% (agarose gel electrophoresis) | assay ≥96% (agarose gel electrophoresis) | assay ≥96% (agarose gel electrophoresis) | assay ≥98% (agarose gel electrophoresis) |
| Quality Level 200 | Quality Level 200 | Quality Level 200 | Quality Level 200 |
| form lyophilized powder | form lyophilized powder | form lyophilized powder | form lyophilized powder |
Still not finding the right product?
Explore all of our products under Bovine Serum Albumin
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Global Trade Item Number
| SKU | GTIN |
|---|---|
| A3733-500G | 04061833498552 |
| A3733-50G | 04061833367872 |
| A3733-10G | 04061833367865 |
| A3733-1KG | 04061826680032 |
| A3733-100G | 04061833367858 |

