Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Bovine Serum Albumin
Synonym(s):
Albumin bovine serum, BSA, Bovine albumin, Albumin, Bovine Serum, 10% Aqueous Solution, Nuclease-Free, OmniPur
®
BSA, Fraction V, Cold Alcohol Isolation, Albumin, Bovine Serum, Cohn Fraction, 30% Aqueous Solution
CAS Number:
9048-46-8
EC Number:
232-936-2
Products
Product Category
albumins (41)
proteins (1)
Brand
Sigma-Aldrich (42)
Biological Source
Feature
pH Value
Physical Form
Product Line
Purity
Quality Segment
Geographic Origin
Package Size
Technique
Filter Products
Showing 1-20 of 42
Page 1 of 3
Page 1 of 3
Compare
Product Number
Description
Pricing
A7030
Bovine Serum Albumin,
heat shock fraction, protease free, fatty acid free, essentially globulin free, pH 7, ≥98%
Expand
View Pricing
A2153
Bovine Serum Albumin,
lyophilized powder, ≥96% (agarose gel electrophoresis)
Expand
View Pricing
A9647
Bovine Serum Albumin,
heat shock fraction, pH 7, ≥98%
Expand
View Pricing
A9418
Bovine Serum Albumin,
lyophilized powder, BioReagent, suitable for cell culture
Expand
View Pricing
A8806
Bovine Serum Albumin,
fatty acid free, low endotoxin, lyophilized powder, BioReagent, suitable for cell culture, ≥96% (agarose gel electrophoresis)
Expand
View Pricing
A3059
Bovine Serum Albumin,
heat shock fraction, protease free, essentially globulin free, pH 7, ≥98%
Expand
View Pricing
A3294
Bovine Serum Albumin,
heat shock fraction, protease free, pH 7, ≥98%
Expand
View Pricing
A4503
Bovine Serum Albumin,
cold ethanol fraction, pH 5.2, ≥96%
Expand
View Pricing
A7906
Bovine Serum Albumin,
heat shock fraction, pH 7, ≥98%
Expand
View Pricing
A6003
Bovine Serum Albumin,
lyophilized powder, essentially fatty acid free, ≥96% (agarose gel electrophoresis)
Expand
View Pricing
A8022
Bovine Serum Albumin,
heat shock fraction, pH 5.2, ≥96%
Expand
View Pricing
A7888
Bovine Serum Albumin,
heat shock fraction, suitable for RIA, pH 5.2, ≥96%
Expand
View Pricing
126615
Bovine Serum Albumin,
10% Aqueous Solution, Nuclease-Free
Expand
View Pricing
A7638
Bovine Serum Albumin,
lyophilized powder, essentially globulin free, ≥99% (agarose gel electrophoresis)
Expand
View Pricing
A3803
Bovine Serum Albumin,
heat shock fraction, lyophilized powder, essentially fatty acid free, ≥98% (agarose gel electrophoresis)
Expand
View Pricing
A1470
Bovine Serum Albumin,
lyophilized powder, low endotoxin, BioReagent, suitable for cell culture, ≥98% (agarose gel electrophoresis)
Expand
View Pricing
05470
Bovine Serum Albumin,
lyophilized powder, crystallized, ≥98.0% (GE)
Expand
View Pricing
B6917
Bovine Serum Albumin,
lyophilized powder, suitable for (for molecular biology), Non-acetylated
Expand
View Pricing
A3311
Bovine Serum Albumin,
powder, BioXtra
Expand
View Pricing
A0281
Bovine Serum Albumin,
lyophilized powder, essentially fatty acid free and essentially globulin free, ≥99% (agarose gel electrophoresis)
Expand
View Pricing
Page 1 of 3
Page 1 of 3