Skip to Content
Merck

Skip To

A3449

Acrylamide/Bis-acrylamide solution

30%, BioReagent, suitable for electrophoresis, 19:1

Synonym(s):

30% Acrylamide/Bisacrylamide 19:1, Polyacrylamide solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
41105319
PubChem Substance ID:
NACRES:
NA.25
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Acrylamide/Bis-acrylamide, 30% solution, BioReagent, suitable for electrophoresis, 19:1

sterility

0.2 μm filtered

Quality Segment

product line

BioReagent

form

liquid

feed ratio

19:1

technique(s)

electrophoresis: suitable

suitability

suitable for electrophoresis

foreign activity

Protease, none detected

storage temp.

2-8°C

SMILES string

NC(=O)C=C.C=CC(=O)NCNC(=O)C=C

InChI

1S/C7H10N2O2.C3H5NO/c1-3-6(10)8-5-9-7(11)4-2;1-2-3(4)5/h3-4H,1-2,5H2,(H,8,10)(H,9,11);2H,1H2,(H2,4,5)

InChI key

OCQZXMCGTAWGEQ-UHFFFAOYSA-N

General description

Polyacrylamide gels are prepared using acrylamide and bis-acrylamide, where bis-acrylamide is a cross-linking agent. On the other hand, acrylamide is a water soluble monomer which helps in forming a transparent stable insoluble gel.

Application

These 30% (w/w) acrylamide/bis-acrylamide solutions are available at the most common ratios for use in protein and nucleic acid electrophoresis. The concentration is based on the total weight of both the acrylamide and bis-acrylamide.
Acrylamide/Bis-acrylamide, 30% solution has been used as a constituent in the preparation of 5% stacking gel and 7% resolving gel for sodium dodecyl sulfate-polyacrylamide gel electrophoresis (SDS-PAGE).

Preparation Note

Solution prepared from electrophoresis grade acrylamide and bis-acrylamide in ultra-pure water.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A3574A9926A2917
form

liquid

form

liquid

form

liquid

form

powder

technique(s)

electrophoresis: suitable

technique(s)

electrophoresis: suitable

technique(s)

electrophoresis: suitable

technique(s)

-

feed ratio

19:1

feed ratio

29:1

feed ratio

19:1

feed ratio

19:1 (ratio)

suitability

suitable for electrophoresis

suitability

suitable for electrophoresis

suitability

suitable for electrophoresis

suitability

-

foreign activity

Protease, none detected

foreign activity

Protease, none detected

foreign activity

-

foreign activity

DNAse (exonuclease), NICKase (endonuclease), RNAse, and protease, none detected

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

-


Still not finding the right product?


pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Eye Irrit. 2 - Muta. 1B - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 Oral

target_organs

Peripheral nervous system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters