Skip to Content
Merck

Skip To

A2917

Acrylamide/Bis-acrylamide

BioReagent, 19:1 (ratio), Molecular Biology

Synonym(s):

Acrylamide/Bisacrylamide, 19:1 (5% crosslinker), Polyacrylamide solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
41105319
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

Molecular Biology

Quality Segment

product line

BioReagent

form

powder

feed ratio

19:1 (ratio)

foreign activity

DNAse (exonuclease), NICKase (endonuclease), RNAse, and protease, none detected

SMILES string

NC(=O)C=C.C=CC(=O)NCNC(=O)C=C

InChI

1S/C7H10N2O2.C3H5NO/c1-3-6(10)8-5-9-7(11)4-2;1-2-3(4)5/h3-4H,1-2,5H2,(H,8,10)(H,9,11);2H,1H2,(H2,4,5)

InChI key

OCQZXMCGTAWGEQ-UHFFFAOYSA-N

General description

Polyacrylamide gels are prepared using acrylamide and bis-acrylamide, where bis-acrylamide is a cross-linking agent. On the other hand, acrylamide is a water soluble monomer which helps in forming a transparent stable insoluble gel.

Application

Ready to reconstitute dry powder blends are accurately pre-blended to produce a 40% (w/v) stock solution for use in protein and nucleic acid electrophoresis. The concentration is based on the total weight of both the acrylamide and bis-acrylamide.
Acrylamide/Bis-acrylamide has been used:
  • in the preparation of urea polyacrylamide gel during a modified cross-linking affinity purification protocol (cCLAP)
  • in microsatellite genotyping and nonfluorescent elastin (ELN) (GT)n polymorphism genotyping
  • in polyacrylamide gel for separation of proteins in SDS-PAGE for western blotting

Features and Benefits

Eliminates the need to weigh toxic acrylamide and bis-acrylamide.

Preparation Note

Reconstitute by slowly adding the indicated amount of distilled water to the container and dissolve contents.
To make up 100 ml of A2917, slowly add 63 ml distilled or deionized water to the container and dissolve contents. Do not divide the solid. This makes 100 ml of a 40% stock solution.
For the 1 Liter size, use 630 mL of water.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A6050A3449A2792
form

powder

form

powder

form

liquid

form

powder

grade

Molecular Biology

grade

Molecular Biology

grade

-

grade

Molecular Biology

feed ratio

19:1 (ratio)

feed ratio

37:1 (ratio)

feed ratio

19:1

feed ratio

29:1 (ratio)

foreign activity

DNAse (exonuclease), NICKase (endonuclease), RNAse, and protease, none detected

foreign activity

DNase, RNase and protease, none detected

foreign activity

Protease, none detected

foreign activity

DNase, RNase and protease, none detected

product line

BioReagent

product line

BioReagent

product line

BioReagent

product line

BioReagent

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200


Still not finding the right product?

Explore all of our products under Acrylamide/Bis-acrylamide


pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Carc. 1B - Eye Irrit. 2 - Muta. 1B - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 Oral

target_organs

Peripheral nervous system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters