This product is a polyclonal antibody. Polyclonal antibodies are heterologous mixtures of IgG isotypes.
Select a Size
| Pack Size | SKU | Availability | Price |
|---|
About This Item
biological source
rabbit
Quality Level
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
species reactivity
rat, Drosophila, wide range
packaging
antibody small pack of 25 μL
technique(s)
dot blot: 1:10,000, immunohistochemistry (formalin-fixed, paraffin-embedded sections): 2.5 μg/mL using rat cerebellum
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
rat ... Gabra1(29705)
General description
GABA (γ-aminobutyric acid) is a multifunctional molecule found in several organisms, from prokaryotes to vertebrates. It is present in non-nervous structures, such as peripheral nervous and endocrine systems. GABA is formed following decarboxylation of glutamic acid by the enzyme glutamic acid decarboxylase (GAD).
Immunogen
Application
Biochem/physiol Actions
Physical form
Preparation Note
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| antibody form affinity isolated antibody | antibody form purified from hybridoma cell culture | antibody form ascites fluid | antibody form IgG fraction of antiserum |
| biological source rabbit | biological source mouse | biological source mouse | biological source rabbit |
| Quality Level 200 | Quality Level 200 | Quality Level 200 | Quality Level 200 |
| conjugate unconjugated | conjugate - | conjugate unconjugated | conjugate unconjugated |
| form buffered aqueous solution | form buffered aqueous solution | form - | form buffered aqueous solution |
| Gene Information rat ... Gabra1(29705) | Gene Information human ... GABRA1(2554) | Gene Information mouse ... Gabra1(14394) | Gene Information human ... GLUL(2752) |
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
10 - Combustible liquids
wgk
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Instructions
Global Trade Item Number
| SKU | GTIN |
|---|---|
| A2052-100UL | 04061833343944 |
| A2052-.2ML | 04061833343937 |
| A2052-25UL | 04061838451606 |
-
What is the source isotype?
1 answer-
Helpful?
-
-
whats the sequence of recognition of the antibody
1 answer-
The peptide sequence for this antibody has not been determined.
Helpful?
-



