The difference between the Type I and Type II Asialofetuin products does not represent any kind of biochemical or functional difference. These indicate different methods of production and purification. The Type II option, product A1908, utilizes neuraminadase in an enzymatic cleavage step. The Type I, product A4781, utilizes a proprietary modification of that described in Spiro, R.G., J. Biol. Chem., 235, 2860 (1960).
Select a Size
| Pack Size | SKU | Availability | Price |
|---|
About This Item
biological source
bovine
Quality Level
type
Type II (Sigma designation)
form
powder
technique(s)
activity assay: suitable, inhibition assay: suitable
impurities
salt, essentially free, ≤1.0% N-acetylneuraminic acid
solubility
H2O: soluble 1.0 mg/mL, clear to hazy, colorless to very faintly yellow
storage temp.
2-8°C
Application
Biochem/physiol Actions
Preparation Note
1 of 1
This Item | |||
|---|---|---|---|
| biological source bovine | biological source bovine (calf) serum | biological source bovine serum (fetal) | biological source bovine serum (fetal) |
| technique(s) activity assay: suitable, inhibition assay: suitable | technique(s) - | technique(s) cell culture | mammalian: suitable | technique(s) HPLC: suitable, MALDI-MS: suitable |
| form powder | form powder | form lyophilized powder | form lyophilized powder |
| storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C |
| impurities salt, essentially free, ≤1.0% N-acetylneuraminic acid | impurities salt, essentially free, ≤0.5% N-acetylneuraminic acid | impurities ≤0.3% free N-acetylneuraminic acid | impurities ~0.2% free N-acetylneuraminic acid |
| Quality Level 200 | Quality Level 300 | Quality Level 200 | Quality Level 200 |
Still not finding the right product?
Explore all of our products under Asialofetuin from fetal calf serum
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| A1908-50MG | 04061833257890 |
| A1908-10MG | 04061831151886 |
| A1908-5MG | 04061831151893 |
-
What is the difference in Type 1 versus Type II asialofetuin?
1 answer-
Helpful?
-



