Skip to Content
Merck

Skip To

A4781

Asialofetuin from fetal calf serum

Type I (Sigma designation)

Synonym(s):

Asialofetuin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
12352202
NACRES:
NA.61
MDL number:
Form:
powder
Biological source:
bovine (calf) serum
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

bovine (calf) serum

Quality Level

type

Type I (Sigma designation)

form

powder

impurities

salt, essentially free, ≤0.5% N-acetylneuraminic acid

solubility

0.85% sodium chloride: soluble 1 mg/mL

storage temp.

2-8°C

General description

Asialofetuin is a glycoprotein with three asparagine-linked triantennary complex carbohydrate chains and terminal N-acetylgalactosamine residues.[1]

Application

Asialofetuin from fetal calf serum has been used:
  • to quantitate the plant Ricin toxin′s B subunit (RTB) lectin activity in β-phaseolin signal peptide (P)–proinsulin gene (INS)–RTB plants by enzyme-linked immunosorbent assay (ELISA)[2]
  • quantitate VP7:RTB fusion protein in transformed potato tissues by ELISA[3]
  • as a glycoprotein substrate to measure the receptor-binding activity of recombinant RTB and NSP490–RTB fusion proteins[4]
  • to study the nature of the interaction between ferritin and the placenta[5]
Incorporation of asialofetuin (AF) in liposomes strongly enhance delivery to and endocytosis by cells displaying the AF receptor, notably hepatocytes. This provides a very efficient route for gene therapy.

Biochem/physiol Actions

Asialofetuin exhibits affinity to asialoglycoprotein receptor (ASGP-R) on hepatocytes and uses the receptor to enter the cell. This property allows asialofetuin to be used as a ligand to deliver drugs to hepatocytes and as a competitive inhibitor to ASGP-R. [1]

Preparation Note

Prepared by a modification of Spiro, R.G., J. Biol. Chem., 235, 2860 (1960).

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
F2379F3004F3385
biological source

bovine (calf) serum

biological source

bovine serum (fetal)

biological source

bovine serum (fetal)

biological source

-

form

powder

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

impurities

salt, essentially free, ≤0.5% N-acetylneuraminic acid

impurities

≤0.3% free N-acetylneuraminic acid

impurities

~0.2% free N-acetylneuraminic acid

impurities

≤0.3% free N-acetylneuraminic acid

Quality Level

300

Quality Level

200

Quality Level

200

Quality Level

200

type

Type I (Sigma designation)

type

-

type

-

type

-


Still not finding the right product?

Explore all of our products under Asialofetuin from fetal calf serum


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Nida Zaidi et al.
International journal of biological macromolecules, 103, 65-73 (2017-05-13)
The SDS-glycoprotein system is mimic of membrane protein-lipid system. Fate of glycoprotein, conformation and the interactive forces involved in membrane milieu are expected to be decided by the net charge on glycoprotein that may change during acidic environment in a
Jakob Meier-Credo et al.
Journal of the American Society for Mass Spectrometry, 33(7), 1293-1302 (2022-06-28)
Identification and sequence determination by mass spectrometry have become routine analyses for soluble proteins. Membrane proteins, however, remain challenging targets due to their hydrophobicity and poor annotation. In particular small membrane proteins often remain unnoticed as they are largely inaccessible
Keiichi Motoyama et al.
Journal of drug delivery, 2011, 476137-476137 (2011-04-15)
The purpose of this study is to evaluate in vitro gene delivery mediated by asialofetuin-appended cationic liposomes (AF-liposomes) associating cyclodextrins (CyD/AF-liposomes) as a hepatocyte-selective nonviral vector. Of various CyDs, AF-liposomes associated with plasmid DNA (pDNA) and γ-cyclodextrin (γ-CyD) (pDNA/γ-CyD/AF-liposomes) showed



Global Trade Item Number

SKUGTIN
A4781-100MG04061831156416
A4781-250MG04061833258187
A4781-50MG04061833375006
A4781-1G04061826740910

Questions

1–2 of 2 Questions  
  1. To wich fetuin corresponds your product A4781 (Asialofetuin from fetal calf serum). Is it Protein Fetuin-A ? Uniprot entry: P12763 FETUA_BOVIN or protein Fetuin-B ? Uniprot entry: Q58D62 FETUB_BOVIN Do you have de amino acid sequence?

    1 answer
    1. The form of fetuin has not been determined. This material has not been sequenced. The minimum allowable sialic acid content is 0.5%. The molecular weight is approximately 44,200 daltons. The starting material for this product has a molecular weight of approximately 48,400 including an estimated 8.7% sialic acid. According to the UniPro values for each, the Fetuin-B molecular weight is calculated at 42,663 while the Fetuin-A is 38,419.

      Helpful?

  2. Hi, What is the difference in Type 1 versus Type II asialofetuin

    1 answer
    1. The difference between the Type I and Type II Asialofetuin products does not represent any kind of biochemical or functional difference. These indicate different methods of production and purification. The Type II option, product A1908, utilizes neuraminadase in an enzymatic cleavage step. The Type I, product A4781, uses a proprietary modification of that described in Spiro, R.G., J. Biol. Chem., 235, 2860 (1960).

      Helpful?

Reviews

No rating value

Active Filters