Skip to Content
Merck

Skip To

A1887

Albumin from human serum

lyophilized powder, essentially fatty acid free

Synonym(s):

HSA

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352202
eCl@ss:
42010201
EC Number:
274-272-6
NACRES:
NA.25
MDL number:
Biological source:
human
Assay:
≥96% (agarose gel electrophoresis)
Form:
lyophilized powder
Technique(s):
ELISA: suitable, tissue culture: suitable, western blot: suitable
Impurities:
HIV I and HIVII, HCV and HBsAg, tested negative, ≤0.007% Fatty Acids
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human

Quality Level

assay

≥96% (agarose gel electrophoresis)

form

lyophilized powder

mol wt

monomer calculated mol wt 66478 Da

technique(s)

ELISA: suitable, tissue culture: suitable, western blot: suitable

impurities

HIV I and HIVII, HCV and HBsAg, tested negative, ≤0.007% Fatty Acids

solubility

H2O: soluble 50 mg/mL, clear to slightly hazy

UniProt accession no.

storage temp.

2-8°C

SMILES string

FC(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C3F8/c4-1(5,2(6,7)8)3(9,10)11

InChI key

QYSGYZVSCZSLHT-UHFFFAOYSA-N

Gene Information

human ... ALB(213)

General description

Albumin is the most abundant plasma protein in human. It has a molecular weight of 66.5 kDa.

Application

Albumin from human serum has been used:
  • as a carrier of CPT-11 (irinotecan, Campto) and SN-38 to investigate its binding to human plasma proteins
  • to study the adsorption of water and acetonitrile (AN) vapor on a human serum albumin
  • to study its effect on wound healing process

Biochem/physiol Actions

Albumins are soluble monomeric proteins found in the body fluids and tissues of animals and in some plant seeds. Serum albumin functions as a carrier protein for steroids, fatty acids, and thyroid hormones. Serum albumins are also vital in regulating the colloidal osmotic pressures of blood.
Serum albumin functions as a carrier protein for steroids, fatty acids, and thyroid hormones, and is vital in regulating the colloidal osmotic pressures of blood. Albumin is also seen to bind to exogenous substances, particularly drugs (e.g., ibuprofen, warfarin), and strongly influence their pharmacokinetics. Oxidative stress leading to changes in the redox state of albumin has widely varied effects on its physiological function.
Serum albumin primarily mediates divalent cation transportation, fatty acid and drug complex formation and transport. The albumin levels in the serum indicate the disease progression in advanced liver disease. Low body weight and decreased serum albumin is associated with colorectal cancer.

Preparation Note

Prepared from albumin (A1653)

Other Notes

View more information on human serum albumin.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A9511A8763A1653
assay

≥96% (agarose gel electrophoresis)

assay

≥97% (agarose gel electrophoresis)

assay

≥99% (agarose gel electrophoresis)

assay

≥96% (agarose gel electrophoresis)

technique(s)

ELISA: suitable, western blot: suitable, tissue culture: suitable

technique(s)

ELISA: suitable, tissue culture: suitable, western blot: suitable

technique(s)

ELISA: suitable, tissue culture: suitable, western blot: suitable

technique(s)

ELISA: suitable, tissue culture: suitable, western blot: suitable

biological source

human

biological source

human

biological source

human

biological source

human

Quality Level

300

Quality Level

300

Quality Level

200

Quality Level

300

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C


Still not finding the right product?

Explore all of our products under Albumin from human serum


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Jasper V Been et al.
Respiratory research, 10, 116-116 (2009-11-26)
Preterm infants are highly susceptible to lung injury. While both chorioamnionitis and antenatal steroids induce lung maturation, chorioamnionitis is also associated with adverse lung development. We investigated the ability of bronchoalveolar lavage fluid (BALF) from ventilated preterm infants to restore
J Barré et al.
Clinical chemistry, 31(1), 60-64 (1985-01-01)
Equilibrium dialysis, ultrafiltration, and ultracentrifugation were compared to determine their reliability and applicability in the study of binding of an anticonvulsant drug, valproic acid, by plasma proteins. We studied drug binding with pooled serum and with solutions of human serum
Di Wu et al.
Scientific reports, 9(1), 1615-1615 (2019-02-09)
The main aim of this work was to gain insight into the binding properties between a food colorant, citrus red 2 (CR), and human serum albumin (HSA), which is the predominant protein in blood plasma. Here, computer simulations and multiple



Global Trade Item Number

SKUGTIN
A1887-100MG04061833343081
A1887-5G04061833343135
A1887-25G04061833343111
A1887-10G04061833343098
A1887-500MG04061833343128
A1887-1G04061833343104

Questions

  1. What is the purpose of the charcoal purification step, and has it been verified that charcoal is not present in the final product? Additionally, how can it be ensured that components of the buffer used for lyophilization are not present in the final product if they are not tested for on the COA?

    1 answer
    1. The purpose of the charcoal purification step is to remove fatty acids from the albumin solution, and it has been verified that no traces of charcoal are present in the final product. As for ensuring that components of the buffer used for lyophilization are not present in the final product, it is currently considered salt-free, but there is potential to characterize it by ICP-MS.

      Helpful?

Reviews

No rating value

Active Filters