Skip to Content
Merck
All Photos(7)

Key Documents

A9511

Sigma-Aldrich

Albumin from human serum

lyophilized powder, ≥97% (agarose gel electrophoresis)

Synonym(s):

HSA

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
eCl@ss:
42010201
NACRES:
NA.25
Pricing and availability is not currently available.

biological source

human

Quality Level

Assay

≥97% (agarose gel electrophoresis)

form

lyophilized powder

mol wt

monomer calculated mol wt 66478 Da

technique(s)

ELISA: suitable
tissue culture: suitable
western blot: suitable

impurities

HIV I and HIVII, HCV and HBsAg, tested negative

solubility

H2O: soluble 50 mg/mL

UniProt accession no.

storage temp.

2-8°C

SMILES string

FC(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C3F8/c4-1(5,2(6,7)8)3(9,10)11

InChI key

QYSGYZVSCZSLHT-UHFFFAOYSA-N

Gene Information

human ... ALB(213)

Looking for similar products? Visit Product Comparison Guide

General description

Albumin is the most copious protein in blood plasma. Liver produces human albumin. [1]
Human albumin possess a molecular weight of 67 kDa.[2]

Application

Albumin from human serum has been used:
  • to standardize the column for molecular-exclusion chromatography[3]
  • to prepare the tobacco mosaic virus (TMV)-poly ethylene glycol (PEG8)-serum albumin (SA) particles[4]
  • to analyze its modifications after oxidation using a Fenton system[5]

Albumin-bound fluorescence was used in serum of patients with chronic renal failure.[3]

Biochem/physiol Actions

Albumin turnover is seen in infants with iron deficiency anemia.[6] Serum albumin is a reliable prognostic indicator in liver disease. Oxidative modification of albumin is seen in advanced liver disease.[7]
Albumins are soluble monomeric proteins found in the body fluids and tissues of animals and in some plant seeds. Serum albumin functions as a carrier protein for steroids, fatty acids, and thyroid hormones. Serum albumins are also vital in regulating the colloidal osmotic pressures of blood.
Serum albumin functions as a carrier protein for steroids, fatty acids, and thyroid hormones, and is vital in regulating the colloidal osmotic pressures of blood. Albumin is also seen to bind to exogenous substances, particularly drugs (e.g., ibuprofen, warfarin), and strongly influence their pharmacokinetics. Oxidative stress leading to changes in the redox state of albumin has widely varied effects on its physiological function.

Preparation Note

Prepared using method IV of Cohn, E.J., et al., J. Am. Chem. Soc., 69, 1753 (1947).

Other Notes

View more information on human serum albumin.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H A Schwertner et al.
Clinical chemistry, 26(5), 649-652 (1980-04-01)
A band that is strongly fluorescent and migrates electrophoretically with serum albumin is commonly found in electrophoretograms of sera from patients with chronic renal failure. We sought to determine whether the fluorescence originates from binding of certain still-unidentified metabolites or
Elham Behzadi et al.
International journal of nanomedicine, 14, 257-270 (2019-01-16)
Recently, nanomaterials have moved into biological and medicinal implementations like cancer therapy. Therefore, before clinical trials, their binding to plasma proteins like human serum albumin (HSA) and their cytotoxic effects against normal and cancer cell lines should be addressed. Herein
Serum albumin camouflage of plant virus based nanoparticles prevents their antibody recognition and enhances pharmacokinetics
Pitek A, et al.
Biomaterials, 89(7-8), 89-97 (2016)
Effects of Fenton Reaction on Human Serum Albumin: An In Vitro Study
Khosravifarsani M, et al.
Electron Physician, 8(9), 2970-2970 (2016)
Daniele Sanna et al.
Inorganic chemistry, 49(1), 174-187 (2009-12-02)
The possible biotransformations in the blood serum of four representative insulin-enhancing vanadium compounds, [VO(6-mepic)(2)], cis-[VO(pic)(2)(H(2)O)], [VO(acac)(2)], and [VO(dhp)(2)], where 6-mepic, pic, acac, and dhp indicate the deprotonated forms of 6-methylpicolinic and picolinic acids, acetylacetone, and 1,2-dimethyl-3-hydroxy-4(1H)-pyridinone, were examined. In particular

Articles

Explore serum albumin's role in serum-free cell culture systems for biomanufacturing.

Explore serum albumin's role in serum-free cell culture systems for biomanufacturing.

Explore serum albumin's role in serum-free cell culture systems for biomanufacturing.

Explore serum albumin's role in serum-free cell culture systems for biomanufacturing.

Questions

1–4 of 4 Questions  
  1. アルブミンの屈折率 情報ありますか?

    1 answer
    1. This product is not tested for refractive index, however, the incremental refractive index is reported in the general Human Serum Albumin datasheet.

      See page 2, PHYSICAL PROPERTIES, at the link below:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/175/446/a9511pis.pdf

      Helpful?

  2. Can Product A9511, Human serum albumin (HSA), be used for cell culture applications?

    1 answer
    1. Although A9511 is not sterile or endotoxin tested, it can probably still be used in cell culture. Filtering a solution of HSA through a 0.45 micron filter, then a 0.2 micron filter would provide a solution that is sterile, and likely suitable for cells. A5843 is an HSA product that has been endotoxin tested.

      Helpful?

  3. What are the impurities for Product A9511, Human serum albumin (HSA)?

    1 answer
    1. The impurities present are alpha-globulin and beta-globulin.

      Helpful?

  4. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service