Skip to Content
Merck

Skip To

08607

2-Ethyl-1-hexanol

analytical standard

Synonym(s):

Isooctyl alcohol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
CH3(CH2)3CH(C2H5)CH2OH
CAS Number:
Molecular Weight:
130.23
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
EC Number:
203-234-3
Beilstein/REAXYS Number:
1719280
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Level

vapor density

4.49 (vs air)

vapor pressure

0.2 mmHg ( 20 °C)

assay

≥99.5% (GC)

autoignition temp.

550 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

0.88 %, 104 °F, 9.7 %, 113 °F

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

refractive index

n20/D 1.431

bp

183-186 °C (lit.)

mp

−76 °C (lit.)

density

0.833 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

SMILES string

CCCCC(CC)CO

InChI

1S/C8H18O/c1-3-5-6-8(4-2)7-9/h8-9H,3-7H2,1-2H3

InChI key

YIWUKEYIRIRTPP-UHFFFAOYSA-N

General description

2-Ethyl-1-hexanol is an industrial chemical widely used in the production of polyvinyl chloride(PVC) plasticizers and is a potent indoor air pollutant emitted mainly from furniture or carpets with a PVC backing.
This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find all available reference materials for compounds listed in 10/2011 here

Application

2-Ethyl-1-hexanol may be used as a reference standard for the determination of 2-Ethyl-1-hexanol in drinking water by gas chromatography with flame ionization detection.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
W315109040508.00990
grade

analytical standard

grade

FG, Fragrance grade, Halal, Kosher

grade

-

grade

synthesis grade

Quality Level

100

Quality Level

300, 400

Quality Level

200

Quality Level

200

bp

183-186 °C (lit.)

bp

183-186 °C (lit.)

bp

183-186 °C (lit.)

bp

184 °C/1013 hPa

assay

≥99.5% (GC)

assay

≥99%

assay

≥99.0% (GC)

assay

≥99% (GC)

refractive index

n20/D 1.431

refractive index

n20/D 1.431 (lit.)

refractive index

n20/D 1.431

refractive index

-

expl. lim.

0.88 %, 104 °F, 9.7 %, 113 °F

expl. lim.

0.88 %, 104 °F, 9.7 %, 113 °F

expl. lim.

0.88 %, 104 °F, 9.7 %, 113 °F

expl. lim.

1.1-12.7 % (v/v)


Still not finding the right product?

Explore all of our products under 2-Ethyl-1-hexanol


pictograms

Exclamation mark

signalword

Warning

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

167.0 °F - closed cup

flash_point_c

75 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



2-Ethyl-1-hexanol in indoor air as a possible cause of sick building symptoms.
Kamijima M, et al.
Journal of Occupational Health, 44(3), 186-191 (2002)
Determination of 2-ethyl-1-hexanol as contaminant in drinking water.
Vitali M, et al.
Journal of AOAC (Association of Official Analytical Chemists) International, 76, 1133-1133 (1993)
D Fragiadakis et al.
The Journal of chemical physics, 132(14), 144505-144505 (2010-04-22)
The dielectric spectra of most simple liquids are characterized by two relaxation processes: (i) the alpha-process, an intense, broad non-Debye relaxation with a non-Arrhenius temperature dependence and (ii) a beta process, evident mainly below the glass transition and having nearly



Global Trade Item Number

SKUGTIN
318256-5G04061837830495
318256-25G04061837830488
08607-5ML-F04061837240164
08607-1ML-F04061838652737

Questions

Reviews

No rating value

Active Filters