Skip to Content
Merck
All Photos(1)

Key Documents

04050

Sigma-Aldrich

2-Ethyl-1-hexanol

puriss., ≥99.0% (GC)

Synonym(s):

Isooctyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CH(C2H5)CH2OH
CAS Number:
Molecular Weight:
130.23
Beilstein:
1719280
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
≥99.0% (GC)
bp:
183-186 °C (lit.)
vapor pressure:
0.2 mmHg ( 20 °C)
Pricing and availability is not currently available.

vapor density

4.49 (vs air)

Quality Level

vapor pressure

0.2 mmHg ( 20 °C)

grade

puriss.

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

550 °F

expl. lim.

0.88 %, 104 °F
9.7 %, 113 °F

refractive index

n20/D 1.431 (lit.)
n20/D 1.431

bp

183-186 °C (lit.)

mp

−76 °C (lit.)

density

0.833 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

CCCCC(CC)CO

InChI

1S/C8H18O/c1-3-5-6-8(4-2)7-9/h8-9H,3-7H2,1-2H3

InChI key

YIWUKEYIRIRTPP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • 2-Ethyl-1-hexanol is widely used in the synthesis of bis(2-ethylhexyl) phthalate (DEHP), a well-known plasticizer.[1][2]
  • It can be used in the ring-opening alcoholysis of epoxidized vegetable ois to prepare bio-lubricants with low pour point.[3]
  • It is employed as an organic solvent for applications such as post-synthetic modification of metal-organic frameworks (MOFs)[4] and esterification reaction of triphenylenes to synthesize liquid crystals.[5]
  • It can also be used as a solvent and in the preparation of nanocrystalline titania thin films by screen printing.[6]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

167.0 °F - closed cup

Flash Point(C)

75 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Green approach for the preparation of biodegradable lubricant base stock from epoxidized vegetable oil.
Lathi P S and Mattiasson B
Applied Catalysis. B, Environmental, 69(3-4), 207-212 (2007)
2-Ethyl-1-hexanol based screen-printed titania thin films for dye-sensitized solar cells.
Tsoukleris D S, et al.
Solar Energy, 79(4), 422-430 (2005)
Esterification of phthalic anhydride with 1-butanol and 2-ethylhexanol catalyzed by heteropolyacids.
Arabi M, et al.
J. Mol. Catal. A: Chem., 200(1-2), 105-110 (2003)
Metal?organic frameworks with precisely designed interior for carbon dioxide capture in the presence of water.
Fracaroli A M, et al.
Journal of the American Chemical Society, 136(25), 8863-8866 (2014)
An Efficient Route to Stable Room?Temperature Liquid?Crystalline Triphenylenes.
Bock H, et al.
European Journal of Organic Chemistry, 2006(13), 2889-2893 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service