Skip to Content
Merck

Skip To

1.02447

Chloroform

for spectroscopy Uvasol®

Synonym(s):

TCM, Trichloromethane, Methane trichloride, Methyl trichloride, Methylidyne trichloride, Trichloromethane

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
CHCl3
CAS Number:
Molecular Weight:
119.38
UNSPSC Code:
12191502
NACRES:
NA.03
EC Index Number:
200-663-8
MDL number:
Beilstein/REAXYS Number:
1731042
Assay:
≥99% (GC)
Technique(s):
UV/Vis spectroscopy: suitable
Bp:
60.5-61.5 °C (lit.)
Vapor pressure:
160 mmHg ( 20 °C)
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor density

4.1 (vs air)

Quality Segment

vapor pressure

160 mmHg ( 20 °C)

assay

≥99% (GC)

form

liquid

potency

695 mg/kg LD50, oral (Rat), >3980 mg/kg LD50, skin (Rabbit)

contains

1% ethanol as stabilizer

technique(s)

UV/Vis spectroscopy: suitable

impurities

≤0.0002 meq/g Acidity, ≤0.0002 meq/g Alkalinity, ≤0.01% Water

evapn. residue

≤0.0002%

color

APHA: ≤10

transmittance

245 nm, ≥15%, 250 nm, ≥50%, 255 nm, ≥60%, 260 nm, ≥85%, 270 nm, ≥98%

refractive index

n20/D 1.445 (lit.)

bp

60.5-61.5 °C (lit.)

mp

−63 °C (lit.)

solubility

8.7 g/L

density

1.492 g/mL at 25 °C (lit.)

UV absorption

λ: 245 nm Amax: ≤0.82, λ: 250 nm Amax: ≤0.30, λ: 255 nm Amax: ≤0.22, λ: 260 nm Amax: ≤0.07, λ: 270 nm Amax: ≤0.01

storage temp.

2-30°C

SMILES string

ClC(Cl)Cl

InChI

1S/CHCl3/c2-1(3)4/h1H

InChI key

HEDRZPFGACZZDS-UHFFFAOYSA-N

General description

Accurate analytic results in UV/VIS and infrared spectroscopy depend on the use of very pure solvents for sample preparation. The Uvasol<TMSYMBOL></TMSYMBOL> solvents range has been specially designed for spectroscopy and other applications requiring solvents of the highest spectral purity. The refinement process allows a greater degree of security in applications and avoids misinterpretation of analytical results caused by traces of UV, IR and fluorescence contamination. Uvasol<TMSYMBOL></TMSYMBOL> solvents offer best UV transmittance. In all specifications the minimum transmittance for 5 typical wavelengths are identified.

Application


  • Preparation and application of stimuli-responsive PET TeMs: Utilizes RAFT graft block copolymerisation of styrene and acrylic acid for the separation of water-oil emulsions, demonstrating the adaptation of chloroform in enhancing polymer properties for environmental applications (Muslimova et al., 2024).

  • Effect of Stool Sampling on a Routine Clinical Method: Describes the use of chloroform in gas chromatography-mass spectrometry for the quantification of short-chain fatty acids in stool samples, highlighting its importance in clinical diagnostics (Mahdi et al., 2024).

  • Improved lipid analysis using a 2D-LC-MS system: Discusses a novel injection procedure involving chloroform for enhanced lipid profiling, showcasing its critical role in advanced biochemical analysis (Pánczél et al., 2024).

  • Ecological Risk Assessment of Three Pesticide Additives in Soil: Addresses the application of chloroform in the remediation of contaminated soils, emphasizing its utility in environmental toxicology and soil science (Li et al., 2024).

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

UVASOL is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.0243236691932211-M
assay

≥99% (GC)

assay

≥99.8% (GC)

assay

≥99.8%

assay

99.0-99.4% (GC)

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

-

technique(s)

UV/Vis spectroscopy: suitable

technique(s)

gas chromatography (GC): suitable

technique(s)

UV/Vis spectroscopy: suitable

technique(s)

RNA extraction: suitable

solubility

8.7 g/L

solubility

8.7 g/L

solubility

-

solubility

-

form

liquid

form

liquid

form

liquid

form

liquid

bp

60.5-61.5 °C (lit.)

bp

60.5-61.5 °C (lit.)

bp

60.5-61.5 °C (lit.)

bp

60.5-61.5 °C (lit.)


Still not finding the right product?

Explore all of our products under Chloroform


pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

wgk

WGK 3

flash_point_f

does not flash

flash_point_c

does not flash

target_organs

Central nervous system, Liver,Kidney

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Global Trade Item Number

SKUGTIN
102447050004022536033716
102447250004022536033730

Questions

Reviews

No rating value

Active Filters