Skip to Content
Merck

Skip To

1.02432

Chloroform

for gas chromatography ECD and FID SupraSolv®

Synonym(s):

TCM, Trichloromethane, Methane trichloride, Methyl trichloride, Methylidyne trichloride, Trichloromethane

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
CHCl3
CAS Number:
Molecular Weight:
119.38
UNSPSC Code:
12191502
NACRES:
NA.03
EC Index Number:
200-663-8
MDL number:
Beilstein/REAXYS Number:
1731042
Assay:
≥99.8% (GC)
Technique(s):
gas chromatography (GC): suitable
Bp:
60.5-61.5 °C (lit.)
Vapor pressure:
160 mmHg ( 20 °C)
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor density

4.1 (vs air)

Quality Level

vapor pressure

160 mmHg ( 20 °C)

product line

SupraSolv®

assay

≥99.8% (GC)

form

liquid

potency

695 mg/kg LD50, oral (Rat), >3980 mg/kg LD50, skin (Rabbit)

contains

Ethanol as stabilizer

technique(s)

gas chromatography (GC): suitable

refractive index

n20/D 1.445 (lit.)

bp

60.5-61.5 °C (lit.)

mp

−63 °C (lit.)

solubility

8.7 g/L

density

1.492 g/mL at 25 °C (lit.)

storage temp.

2-30°C

SMILES string

ClC(Cl)Cl

InChI

1S/CHCl3/c2-1(3)4/h1H

InChI key

HEDRZPFGACZZDS-UHFFFAOYSA-N

General description

SupraSolvECD and FID is specially developed for gas chromatography in combination with ECD (Electron Capture Detector) and FID (Flame Ionization Detector). SupraSolv solvents offer the largest specified retention time range, a clear baseline, and a minimal signal-to-noise ratio, leading to reliable and reproducible analysis results.

Application

Chloroformcan be used as a solvent in the determination of peroxide value.

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

SUPRASOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.024451.02447CX1055
technique(s)

gas chromatography (GC): suitable

technique(s)

-

technique(s)

UV/Vis spectroscopy: suitable

technique(s)

-

assay

≥99.8% (GC)

assay

≥99.8%, 90-99.4% (GC)

assay

≥99% (GC)

assay

≥99.8% chloroform + ethanol basis

Quality Level

100

Quality Level

300

Quality Level

100

Quality Level

200

solubility

8.7 g/L

solubility

-

solubility

8.7 g/L

solubility

8.7 g/L

form

liquid

form

liquid

form

liquid

form

liquid

mp

−63 °C (lit.)

mp

−63 °C (lit.)

mp

−63 °C (lit.)

mp

−63 °C (lit.)


Still not finding the right product?

Explore all of our products under Chloroform


pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

target_organs

Central nervous system, Liver,Kidney

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

flash_point_f

does not flash

flash_point_c

does not flash



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Extending high fatty foods shelf-life protecting from lipid oxidation and microbiological contamination: an approach using active packaging with pomegranate extract
Andrade MA, et al.
Coatings, 13(1), 93-93 (2023)



Questions

Reviews

No rating value

Active Filters